Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Mouse TECK ELISA Kit

Mouse TECK ELISA Kit

Mouse TECK ELISA Kit, 96 tests
Katalog # ABCAAB100743-96
Leverandør:   Abcam
Mouse TECK ELISA Kit
Katalog # ABCAAB100743-96
Leverandør:  Abcam
CAS nummer:  
Mouse TECK ELISA Kit
Mouse TECK ELISA Kit

Mouse TECK ELISA Kit, 96 tests

CAS nummer:  
Price
6.390,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Mouse TECK ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse TECK in Mouse in plasma, cell culture supernatant, serum samples.

Specifikationer

Specifikationer

  • Beskrivelse:
    Mouse TECK ELISA Kit
  • Test pr. sæt:
    96 tests
  • Cat. No.:
    ABCAAB100743-96
  • Sensitivitet:
    15 pg/ml
  • Format:
    Pre-coated
  • Assay varighed:
    Multiple steps
  • Primary antibody reactivity:
    Mouse
  • Detektionsområde:
    20,5 - 2000 pg/ml
  • Target protein:
    TECK
  • Detektionsmetode:
    Colorimetric
  • Prøvetype:
    Plasma, Cell culture supernatant, Serum
  • Opbevaringstemperatur:
    4 °C
  • Regulatory status:
    RUO
  • Analysetype:
    Sandwich

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.