Mouse IgM ELISA Kit
Mouse IgM ELISA kit, 96 tests
ABCAAB133047-96
: Abcam
Mouse IgM ELISA kit ab133047 is used to quantify Mouse IgM with a sensitivity of 0,6 ng/ml. Sandwich ELISA kit.
- Beskrivelse:Mouse IgM ELISA kit
- Test pr. sæt:96 tests
- Sensitivitet:0,6 ng/ml
- Assay varighed:Multiple steps
- Primary antibody reactivity:Mouse
- Detektionsområde:3,91 - 250 ng/ml
- Prøvetype:Cell culture supernatant, Serum, Ascites Fluid
- Time to Results:1 h 30 min
- Analysetype:Sandwich
- Cat. No.:ABCAAB133047-96
- Format:Pre-coated
- Target protein:IgM
- Detektionsmetode:Colorimetric
- Opbevaringstemperatur:4 °C
- Regulatory status:RUO
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



