Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Factor X ELISA Kit

Human Factor X ELISA Kit

Human Factor X ELISA kit, 96 tests
Katalog # ABCAAB108832-96
Leverandør:   Abcam
Human Factor X ELISA Kit
Katalog # ABCAAB108832-96
Leverandør:  Abcam
CAS nummer:  
Human Factor X ELISA Kit
Human Factor X ELISA Kit

Human Factor X ELISA kit, 96 tests

CAS nummer:  
Price
6.150,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Human Factor X ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Factor X in Human in Cerebral Spinal Fluid, Saliva, Urine, Plasma, Milk, Serum samples.

Specifikationer

Specifikationer

  • Beskrivelse:
    Human Factor X ELISA kit
  • Test pr. sæt:
    96 tests
  • Sensitivitet:
    0,16 ng/ml
  • Assay varighed:
    Multiple steps
  • Primary antibody reactivity:
    Human
  • Detektionsområde:
    0,781 - 50 ng/ml
  • Prøvetype:
    Cerebral Spinal Fluid, Saliva, Urine, Plasma, Milk, Serum
  • Time to Results:
    4 h 0 min
  • Analysetype:
    Sandwich
  • Cat. No.:
    ABCAAB108832-96
  • Format:
    Pre-coated
  • Target protein:
    Factor X
  • Detektionsmetode:
    Colorimetric
  • Opbevaringstemperatur:
    +4 °C
  • Regulatory status:
    RUO

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.