Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Factor B ELISA Kit

Human Factor B ELISA Kit

Human Factor B ELISA kit, 96 tests
Katalog # ABCAAB137973-96
Leverandør:   Abcam
Human Factor B ELISA Kit
Katalog # ABCAAB137973-96
Leverandør:  Abcam
CAS nummer:  
Human Factor B ELISA Kit
Human Factor B ELISA Kit

Human Factor B ELISA kit, 96 tests

CAS nummer:  
Price
6.150,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Human Factor B ELISA Kit ab137973 is used to quantify Human Factor B with a sensitivity of 39 pg/ml. Sandwich ELISA Kit.

Specifikationer

Specifikationer

  • Beskrivelse:
    Human Factor B ELISA kit
  • Test pr. sæt:
    96 tests
  • Sensitivitet:
    0,98 ng/ml
  • Assay varighed:
    Multiple steps
  • Primary antibody reactivity:
    Human
  • Detektionsområde:
    2,188 - 140 ng/ml
  • Prøvetype:
    Milk, Plasma, Saliva, Serum, Cerebral Spinal Fluid
  • Time to Results:
    4 h 0 min
  • Analysetype:
    Sandwich
  • Cat. No.:
    ABCAAB137973-96
  • Format:
    Pre-coated
  • Target protein:
    Factor B
  • Detektionsmetode:
    Colorimetric
  • Opbevaringstemperatur:
    +4 °C
  • Regulatory status:
    RUO

Specifikationer