Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Mouse TRAIL ELISA Kit

Mouse TRAIL ELISA Kit

Mouse TRAIL ELISA Kit, 96 tests
Katalog # ABCAAB193730-96
Leverandør:   Abcam
Mouse TRAIL ELISA Kit
Katalog # ABCAAB193730-96
Leverandør:  Abcam
CAS nummer:  
Mouse TRAIL ELISA Kit
Mouse TRAIL ELISA Kit

Mouse TRAIL ELISA Kit, 96 tests

CAS nummer:  
Price
6.150,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Mouse TRAIL ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse TRAIL in Mouse in plasma, cell culture supernatant, serum samples.

Specifikationer

Specifikationer

  • Beskrivelse:
    Mouse TRAIL ELISA Kit
  • Test pr. sæt:
    96 tests
  • Cat. No.:
    ABCAAB193730-96
  • Sensitivitet:
    2 pg/ml
  • Format:
    Pre-coated
  • Assay varighed:
    Multiple steps
  • Primary antibody reactivity:
    Mouse
  • Detektionsområde:
    2 - 600 pg/ml
  • Target protein:
    TRAIL
  • Detektionsmetode:
    Colorimetric
  • Prøvetype:
    Plasma, Cell culture supernatant, Serum
  • Opbevaringstemperatur:
    4 °C
  • Regulatory status:
    RUO
  • Analysetype:
    Sandwich

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.