Mouse ECM1 ELISA Kit
Mouse ECM1 ELISA Kit, 96 tests
ABCAAB252368-1X96
: Abcam
Mouse ECM1 ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse ECM1 with a sensitivity of 19 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.
- Beskrivelse:Mouse ECM1 ELISA Kit
- Test pr. sæt:96 tests
- Sensitivitet:19 pg/ml
- Assay varighed:One step
- Primary antibody reactivity:Mouse
- Detektionsområde:125 - 8000 pg/ml
- Prøvetype:Heparin Plasma, Citrate plasma, Serum, EDTA Plasma
- Time to Results:1 h 30 min
- Analysetype:Sandwich
- Cat. No.:ABCAAB252368-1X96
- Format:Pre-coated
- Target protein:Extracellular matrix protein 1
- Detektionsmetode:Colorimetric
- Opbevaringstemperatur:4 °C
- Regulatory status:RUO



