Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Mouse CD137 ELISA Kit

Mouse CD137 ELISA Kit

Mouse CD137 ELISA kit, 96 tests
Katalog # ABCAAB205577-96
Leverandør:   Abcam
Mouse CD137 ELISA Kit
Katalog # ABCAAB205577-96
Leverandør:  Abcam
CAS nummer:  
Mouse CD137 ELISA Kit
Mouse CD137 ELISA Kit

Mouse CD137 ELISA kit, 96 tests

CAS nummer:  
Price
5.490,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Mouse CD137 ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse CD137 with a sensitivity of 14 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.

Specifikationer

Specifikationer

  • Beskrivelse:
    Mouse CD137 ELISA kit
  • Test pr. sæt:
    96 tests
  • Sensitivitet:
    14 pg/ml
  • Assay varighed:
    One step
  • Primary antibody reactivity:
    Mouse
  • Detektionsområde:
    78,125 - 5000 pg/ml
  • Prøvetype:
    Plasma, Cell culture extracts, Tissue Extracts, Cell culture supernatant, Serum
  • Time to Results:
    1 h 30 min
  • Analysetype:
    Sandwich
  • Cat. No.:
    ABCAAB205577-96
  • Format:
    Pre-coated
  • Target protein:
    CD137
  • Detektionsmetode:
    Colorimetric
  • Opbevaringstemperatur:
    +4 °C
  • Regulatory status:
    RUO

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.