Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Mouse PSAP ELISA Kit

Mouse PSAP ELISA Kit

Mouse PSAP ELISA kit, 96 tests
Katalog # ABCAAB277399-1X96
Leverandør:   Abcam
Mouse PSAP ELISA Kit
Katalog # ABCAAB277399-1X96
Leverandør:  Abcam
CAS nummer:  
Mouse PSAP ELISA Kit
Mouse PSAP ELISA Kit

Mouse PSAP ELISA kit, 96 tests

CAS nummer:  
Price
6.000,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Mouse PSAP ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse PSAP in Mouse in Plasma, Cell culture supernatant, Serum samples.

Specifikationer

Specifikationer

  • Beskrivelse:
    Mouse PSAP ELISA kit
  • Test pr. sæt:
    96 tests
  • Cat. No.:
    ABCAAB277399-1X96
  • Sensitivitet:
    14 pg/ml
  • Format:
    Pre-coated
  • Assay varighed:
    Multiple steps
  • Primary antibody reactivity:
    Mouse
  • Detektionsområde:
    12,29 - 25000 pg/ml
  • Target protein:
    PSAP
  • Detektionsmetode:
    Colorimetric
  • Prøvetype:
    Plasma, Cell culture supernatant, Serum
  • Opbevaringstemperatur:
    +4 °C
  • Regulatory status:
    RUO
  • Analysetype:
    Sandwich

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.