Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human PYK2 ELISA Kit

Human PYK2 ELISA Kit

Human PYK2 ELISA Kit, 96 tests
Katalog # ABCAAB226892-96
Leverandør:   Abcam
Human PYK2 ELISA Kit
Katalog # ABCAAB226892-96
Leverandør:  Abcam
CAS nummer:  
Human PYK2 ELISA Kit
Human PYK2 ELISA Kit

Human PYK2 ELISA Kit, 96 tests

CAS nummer:  
Price
5.760,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Human PYK2 ELISA kit is a single-wash 90-min Simplestep used to quantify Human PYK2 with a sensitivity of 11.2 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.

Specifikationer

Specifikationer

  • Beskrivelse:
    Human PYK2 ELISA Kit
  • Test pr. sæt:
    96 tests
  • Sensitivitet:
    11,2 pg/ml
  • Assay varighed:
    One step
  • Primary antibody reactivity:
    Human
  • Detektionsområde:
    78 - 5000 pg/ml
  • Prøvetype:
    Cell culture extracts, Tissue Extracts
  • Time to Results:
    1 hr 30 min
  • Analysetype:
    Sandwich
  • Cat. No.:
    ABCAAB226892-96
  • Format:
    Pre-coated
  • Target protein:
    PYK2
  • Detektionsmetode:
    Colorimetric
  • Opbevaringstemperatur:
    4 °C
  • Regulatory status:
    RUO

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.