Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human CD40 ELISA Kit

Human CD40 ELISA Kit

Human CD40 ELISA Kit, 96 tests
Katalog # ABCAAB99990-96
Leverandør:   Abcam
Human CD40 ELISA Kit
Katalog # ABCAAB99990-96
Leverandør:  Abcam
CAS nummer:  
Human CD40 ELISA Kit
Human CD40 ELISA Kit

Human CD40 ELISA Kit, 96 tests

CAS nummer:  
Price
6.390,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Human CD40 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human CD40 in Human in Plasma, Cell culture supernatant, Serum samples.

Specifikationer

Specifikationer

  • Beskrivelse:
    Human CD40 ELISA Kit
  • Test pr. sæt:
    96 tests
  • Cat. No.:
    ABCAAB99990-96
  • Sensitivitet:
    50 pg/ml
  • Format:
    Pre-coated
  • Assay varighed:
    Multiple steps
  • Primary antibody reactivity:
    Human
  • Detektionsområde:
    54,87 - 40000 pg/ml
  • Target protein:
    CD40
  • Detektionsmetode:
    Colorimetric
  • Prøvetype:
    Plasma, Cell culture supernatant, Serum
  • Opbevaringstemperatur:
    4 °C
  • Regulatory status:
    RUO
  • Analysetype:
    Sandwich

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.