Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human CD4 ELISA Kit

Human CD4 ELISA Kit

Human CD4 ELISA kit, 96 tests
Katalog # ABCAAB234569-96
Leverandør:   Abcam
Human CD4 ELISA Kit
Katalog # ABCAAB234569-96
Leverandør:  Abcam
CAS nummer:  
Human CD4 ELISA Kit
Human CD4 ELISA Kit

Human CD4 ELISA kit, 96 tests

CAS nummer:  
Price
6.350,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Human CD4 ELISA kit is a single-wash 90-min Simplestep used to quantify Human CD4 with a sensitivity of 13.7 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.

Specifikationer

Specifikationer

  • Beskrivelse:
    Human CD4 ELISA kit
  • Test pr. sæt:
    96 tests
  • Sensitivitet:
    13,7 pg/ml
  • Assay varighed:
    One step
  • Primary antibody reactivity:
    Human
  • Detektionsområde:
    0,469 - 30 ng/ml
  • Prøvetype:
    Cell culture extracts
  • Time to Results:
    1 h 30 min
  • Analysetype:
    Sandwich
  • Cat. No.:
    ABCAAB234569-96
  • Format:
    Pre-coated
  • Target protein:
    CD4
  • Detektionsmetode:
    Colorimetric
  • Opbevaringstemperatur:
    4 °C
  • Regulatory status:
    RUO

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.