Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Human MEF2A ELISA Kit

Human MEF2A ELISA Kit, 96 tests
  ABCAAB314602-96TST
New Product
 :   Abcam
Human MEF2A ELISA Kit
  ABCAAB314602-96TST
 :  Abcam
New Product
 :  
Human MEF2A ELISA Kit
Human MEF2A ELISA Kit

Human MEF2A ELISA Kit, 96 tests

 :  
 
5.920,00 kr
   
1 * 1 Each
 

Human MEF2A ELISA kit is a single-wash 90-min Simplestep used to quantify Human MEF2A with a sensitivity of 0,055 ng/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.

 

 

  • Beskrivelse:
    Human MEF2A ELISA Kit
  • Test pr. sæt:
    96 tests
  • Sensitivitet:
    0,055 ng/ml
  • Assay varighed:
    One step
  • Primary antibody reactivity:
    Human
  • Detektionsområde:
    0,188 - 12 ng/ml
  • Prøvetype:
    Tissue Extracts, Cell Lysate
  • Time to Results:
    1 hr 30 min
  • Analysetype:
    Sandwich
  • Cat. No.:
    ABCAAB314602-96TST
  • Format:
    Pre-coated
  • Target protein:
    MEF2A
  • Detektionsmetode:
    Colorimetric
  • Opbevaringstemperatur:
    4 °C
  • Regulatory status:
    RUO

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.