Human TNSF18/GITRL ELISA Kit
Human TNSF18/GITRL ELISA kit, 96 tests
Katalog # ABCAAB300314-96
Marketsource
Leverandør: Abcam
Gå til:
Product Details & Documents
Human TNSF18/GITRL ELISA kit is a single-wash 90-min Simplestep used to quantify Human TNSF18/GITRL with a sensitivity of 3,276 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair.
Specifikationer
Specifikationer
- Beskrivelse:Human TNSF18/GITRL ELISA kit
- Test pr. sæt:96 tests
- Sensitivitet:3,276 pg/ml
- Assay varighed:One step
- Primary antibody reactivity:Human
- Detektionsområde:12,5 - 800 pg/ml
- Prøvetype:Cell culture supernatant, Citrate plasma, EDTA Plasma, Heparin Plasma, Serum
- Time to Results:1 h 30 min
- Analysetype:Sandwich
- Cat. No.:ABCAAB300314-96
- Format:Pre-coated
- Target protein:TNFSF18/GITRL
- Detektionsmetode:Colorimetric
- Opbevaringstemperatur:4 °C
- Regulatory status:RUO
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



