Orders LinkContactUsLinkComponent
Mouse Interferon alpha 2 ELISA Kit
Mouse Interferon alpha 2 ELISA Kit
Mouse Interferon alpha 2 ELISA Kit, 96 tests
Katalog # ABCAAB211648-96
Leverandør:   Abcam
Mouse Interferon alpha 2 ELISA Kit
Katalog # ABCAAB211648-96
Leverandør:  Abcam
CAS nummer:  
Mouse Interferon alpha 2 ELISA Kit
Mouse Interferon alpha 2 ELISA Kit
Mouse Interferon alpha 2 ELISA Kit, 96 tests
CAS nummer:  
Price
6.110,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Mouse Interferon alpha 2 ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse Interferon alpha 2 with a sensitivity of 0,9 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.
Specifikationer
Specifikationer
  • Beskrivelse:
    Mouse Interferon alpha 2 ELISA Kit
  • Test pr. sæt:
    96 tests
  • Sensitivitet:
    0,9 pg/ml
  • Assay varighed:
    One step
  • Primary antibody reactivity:
    Mouse
  • Detektionsområde:
    15,36 - 1000 pg/ml
  • Prøvetype:
    Plasma, Cell culture extracts, Tissue Extracts, Cell culture supernatant, Serum
  • Time to Results:
    1 h 30 min
  • Analysetype:
    Sandwich
  • Cat. No.:
    ABCAAB211648-96
  • Format:
    Pre-coated
  • Target protein:
    Interferon alpha 2
  • Detektionsmetode:
    Colorimetric
  • Opbevaringstemperatur:
    4 °C
  • Regulatory status:
    RUO