Human Complement C4 ELISA Kit
Human Complement C4 ELISA Kit, 96 tests
Katalog # ABCAAB108824-96
Leverandør: Abcam
Gå til:
Product Details & Documents
Human Complement C4 ELISA kit is a Competitive ELISA kit for the measurement of Human Complement C4 in Human in plasma, serum samples.
Specifikationer
Specifikationer
- Beskrivelse:Human Complement C4 ELISA Kit
- Test pr. sæt:96 tests
- Sensitivitet:0,53 µg/ml
- Assay varighed:Multiple steps
- Primary antibody reactivity:Human
- Detektionsområde:0,313 - 5 µg/ml
- Prøvetype:Plasma, Serum
- Time to Results:3 hr 0 min
- Analysetype:Competitive
- Cat. No.:ABCAAB108824-96
- Format:Pre-coated
- Target protein:C4
- Detektionsmetode:Colorimetric
- Opbevaringstemperatur:4 °C
- Regulatory status:RUO
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



