Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Human BMP2 ELISA Kit

Human BMP2 ELISA kit, 96 tests
Katalog # ABCAAB277085-1X96
Leverandør:   Abcam
Human BMP2 ELISA Kit
Katalog # ABCAAB277085-1X96
Leverandør:  Abcam
CAS nummer:  
Human BMP2 ELISA Kit
Human BMP2 ELISA Kit

Human BMP2 ELISA kit, 96 tests

CAS nummer:  
Price
5.920,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Human BMP2 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human BMP2 with a sensitivity of 79,8 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.

Specifikationer

Specifikationer

  • Beskrivelse:
    Human BMP2 ELISA kit
  • Test pr. sæt:
    96 tests
  • Sensitivitet:
    79,8 pg/ml
  • Assay varighed:
    One step
  • Primary antibody reactivity:
    Human
  • Detektionsområde:
    187,5 - 12000 pg/ml
  • Prøvetype:
    Cell culture media, heparin plasma, citrate plasma, serum, EDTA plasma
  • Time to Results:
    1 h 30 min
  • Analysetype:
    Sandwich
  • Cat. No.:
    ABCAAB277085-1X96
  • Format:
    Pre-coated
  • Target protein:
    BMP2
  • Detektionsmetode:
    Colorimetric
  • Opbevaringstemperatur:
    4 °C
  • Regulatory status:
    RUO

Specifikationer