Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Mouse IGF1 ELISA Kit

Mouse IGF1 ELISA Kit

Mouse IGF1 ELISA kit, 96 tests
Catalog # ABCAAB100695-96
Supplier:   Abcam
Mouse IGF1 ELISA Kit
Catalog # ABCAAB100695-96
Supplier:  Abcam
CAS Number:  
Mouse IGF1 ELISA Kit
Mouse IGF1 ELISA Kit

Mouse IGF1 ELISA kit, 96 tests

CAS Number:  
Price
6.390,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Mouse IGF1 ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of Mouse IGF1 in Mouse in tissue lysate, plasma, cell culture supernatant, serum samples.

Specifikationer

Specifikationer

  • Beskrivelse:
    Mouse IGF1 ELISA kit
  • Test pr. sæt:
    96 tests
  • Cat. No.:
    ABCAAB100695-96
  • Sensitivitet:
    4 pg/ml
  • Format:
    Pre-coated
  • Assay varighed:
    Multiple steps
  • Primary antibody reactivity:
    Mouse
  • Detektionsområde:
    2,74 - 2000 pg/ml
  • Target protein:
    IGF1
  • Detektionsmetode:
    Colorimetric
  • Prøvetype:
    Tissue Lysate, Plasma, Cell culture supernatant, Serum
  • Opbevaringstemperatur:
    4 °C
  • Regulatory status:
    RUO
  • Analysetype:
    Sandwich

Specifikationer