Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Mouse Apolipoprotein E ELISA Kit

Mouse Apolipoprotein E ELISA kit, 96 tests
Katalog # ABCAAB215086-96
Leverandør:   Abcam
Mouse Apolipoprotein E ELISA Kit
Katalog # ABCAAB215086-96
Leverandør:  Abcam
CAS nummer:  
Mouse Apolipoprotein E ELISA Kit
Mouse Apolipoprotein E ELISA Kit

Mouse Apolipoprotein E ELISA kit, 96 tests

CAS nummer:  
Price
6.080,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Mouse Apolipoprotein E ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse Apolipoprotein E with a sensitivity of 33,9 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair - Cited in over 10 citations.

Specifikationer

Specifikationer

  • Beskrivelse:
    Mouse Apolipoprotein E ELISA kit
  • Test pr. sæt:
    96 tests
  • Sensitivitet:
    33,9 pg/ml
  • Assay varighed:
    One step
  • Primary antibody reactivity:
    Mouse
  • Detektionsområde:
    156,3 - 10000 pg/ml
  • Prøvetype:
    Heparin Plasma, Citrate plasma, Serum, EDTA Plasma
  • Time to Results:
    1 h 30 min
  • Analysetype:
    Sandwich
  • Cat. No.:
    ABCAAB215086-96
  • Format:
    Pre-coated
  • Target protein:
    Apolipoprotein E
  • Detektionsmetode:
    Colorimetric
  • Opbevaringstemperatur:
    4 °C
  • Regulatory status:
    RUO

Specifikationer