Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human COMP ELISA Kit

Human COMP ELISA Kit

Human COMP ELISA kit, 96 tests
Katalog # ABCAAB213764-96
Leverandør:   Abcam
Human COMP ELISA Kit
Katalog # ABCAAB213764-96
Leverandør:  Abcam
CAS nummer:  
Human COMP ELISA Kit
Human COMP ELISA Kit

Human COMP ELISA kit, 96 tests

CAS nummer:  
Price
5.960,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Human COMP ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human COMP in Human in Cell Lysate, Cell culture supernatant, EDTA Plasma, Heparin Plasma, Serum samples.

Specifikationer

Specifikationer

  • Beskrivelse:
    Human COMP ELISA kit
  • Test pr. sæt:
    96 tests
  • Sensitivitet:
    10 pg/ml
  • Assay varighed:
    Multiple steps
  • Primary antibody reactivity:
    Human
  • Detektionsområde:
    156 - 10000 pg/ml
  • Prøvetype:
    Cell Lysate, Cell culture supernatant, EDTA Plasma, Heparin Plasma, Serum
  • Time to Results:
    3 h 30 min
  • Analysetype:
    Sandwich
  • Cat. No.:
    ABCAAB213764-96
  • Format:
    Pre-coated
  • Target protein:
    COMP/Cartilage oligomeric matrix protein
  • Detektionsmetode:
    Colorimetric
  • Opbevaringstemperatur:
    +4 °C
  • Regulatory status:
    RUO

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.