Human CD38 ELISA Kit
Human CD38 ELISA kit, 96 tests
Katalog # ABCAAB267801-1X96
Leverandør: Abcam
Gå til:
Product Details & Documents
Human CD38 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human CD38 with a sensitivity of 7,44 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.
Specifikationer
Specifikationer
- Beskrivelse:Human CD38 ELISA kit
- Test pr. sæt:96 tests
- Sensitivitet:7,44 pg/ml
- Assay varighed:One step
- Primary antibody reactivity:Human
- Detektionsområde:31,25 - 2000 pg/ml
- Prøvetype:Heparin Plasma, Citrate plasma, Cell culture supernatant, Serum, Cell Lysate, EDTA Plasma
- Time to Results:1 h 30 min
- Analysetype:Sandwich
- Cat. No.:ABCAAB267801-1X96
- Format:Pre-coated
- Target protein:CD38
- Detektionsmetode:Colorimetric
- Opbevaringstemperatur:+4 °C
- Regulatory status:RUO



