Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Complement C3a des Arg ELISA Kit

Human Complement C3a des Arg ELISA Kit

Human Complement C3a des Arg ELISA kit, 96 tests
Katalog # ABCAAB133037-96
Leverandør:   Abcam
Human Complement C3a des Arg ELISA Kit
Katalog # ABCAAB133037-96
Leverandør:  Abcam
CAS nummer:  
Human Complement C3a des Arg ELISA Kit
Human Complement C3a des Arg ELISA Kit

Human Complement C3a des Arg ELISA kit, 96 tests

CAS nummer:  
Price
8.230,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Human Complement C3a des Arg ELISA Kit is a Competitive ELISA kit for the measurement of Human Complement C3a des Arg in Human in Plasma samples.

Specifikationer

Specifikationer

  • Beskrivelse:
    Human Complement C3a des Arg ELISA kit
  • Test pr. sæt:
    96 tests
  • Sensitivitet:
    0,12 ng/ml
  • Assay varighed:
    Multiple steps
  • Primary antibody reactivity:
    Human
  • Detektionsområde:
    0,313 - 20 ng/ml
  • Prøvetype:
    Plasma
  • Time to Results:
    3 h 0 min
  • Analysetype:
    Competitive
  • Cat. No.:
    ABCAAB133037-96
  • Format:
    Pre-coated
  • Target protein:
    Complement C3a des Arg
  • Detektionsmetode:
    Colorimetric
  • Opbevaringstemperatur:
    +4 °C
  • Regulatory status:
    RUO

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.