Human Recombinant MAP2 Projection P1
Leverandør: Antibodies.com
Om denne vare
Partial length Human MAP2 Projection P1 suitable for SDS-PAGE, ELISA, Immunogen Grade.
Advarsel: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
2 tilgængelige muligheder nedenfor
Product Details & Documents
Partial length Human MAP2 Projection P1 suitable for SDS-PAGE, ELISA, Immunogen Grade.
Advarsel: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Specifications for Product Family
Specifikationer
Specifikationer
- Catalog No:
- ANTIA270588-25
- ANTIA270588-50
- Protein/peptide name:
- MAP2 Projection P1
- MAP2 Projection P1
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Konjugation:
- Unconjugated
- Unconjugated
- Gen ID:
- UniprotID# P11137
- UniprotID# P11137
- Sekvens:
- DLLTASKMEFHDQQELTPSTAEPSDQKEKESEKQSKPGEDLKHAALVSQPETTKTYPDKKDMQGTEEEKAPLALFGHTLVASLEDMKQKTEPSLVVPGIDLPKEPPTPKEQKDWFIEMPTEAKKDEWGLVAPISPGPLTPMREKDVFDDIPKWEGKQFDSPMPSPFQGGSFTLPLDVMKNEIVTETSPFAPAFLQPDDKKSLQQTSGPATAKDSFKIEEPHEAKPDKMAEAPPSEAMTLPKDAHIPVVEEHVMGKVLEEEKEAINQETVQQRDTFTPSGQEPILTEKETELKLEEKTTISDKEAVPKESKPPKPADEEIGIIQTSTEHTFSEQKDQEPTTDMLKQDSFPVSLEQAVTDSAMTSKTLEKAMTEPSALIEKSSIQELFEMRVDDKDKIEGVGAATSAELDMPFYEDKSGMSKYFETSALKEEATKSIEPGSDYYELSDTRE
- DLLTASKMEFHDQQELTPSTAEPSDQKEKESEKQSKPGEDLKHAALVSQPETTKTYPDKKDMQGTEEEKAPLALFGHTLVASLEDMKQKTEPSLVVPGIDLPKEPPTPKEQKDWFIEMPTEAKKDEWGLVAPISPGPLTPMREKDVFDDIPKWEGKQFDSPMPSPFQGGSFTLPLDVMKNEIVTETSPFAPAFLQPDDKKSLQQTSGPATAKDSFKIEEPHEAKPDKMAEAPPSEAMTLPKDAHIPVVEEHVMGKVLEEEKEAINQETVQQRDTFTPSGQEPILTEKETELKLEEKTTISDKEAVPKESKPPKPADEEIGIIQTSTEHTFSEQKDQEPTTDMLKQDSFPVSLEQAVTDSAMTSKTLEKAMTEPSALIEKSSIQELFEMRVDDKDKIEGVGAATSAELDMPFYEDKSGMSKYFETSALKEEATKSIEPGSDYYELSDTRE
- Bærerfri:
- N
- N
- Formulering:
- Supplied in 6 M Urea.
- Supplied in 6 M Urea.
- Molekylvægt:
- 50 kDa (75 kDa by SDS-PAGE)
- 50 kDa (75 kDa by SDS-PAGE)
- Shipping temperature:
- Shipped on blue ice at 4 °C.
- Shipped on blue ice at 4 °C.
- Tested applications:
- SDS-PAGE, ELISA, Immunogen Grade
- SDS-PAGE, ELISA, Immunogen Grade
Specifikationer
Product Family Options
Produktinformation
- PakningTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.ANTIA270588-25Protein/peptide nameMAP2 Projection P1Protein/peptide typeRecombinantSpeciesHumanKonjugationUnconjugatedGen IDUniprotID# P11137SekvensDLLTASKMEFHDQQELTPSTAEPSDQKEKESEKQSKPGEDLKHAALVSQPETTKTYPDKKDMQGTEEEKAPLALFGHTLVASLEDMKQKTEPSLVVPGIDLPKEPPTPKEQKDWFIEMPTEAKKDEWGLVAPISPGPLTPMREKDVFDDIPKWEGKQFDSPMPSPFQGGSFTLPLDVMKNEIVTETSPFAPAFLQPDDKKSLQQTSGPATAKDSFKIEEPHEAKPDKMAEAPPSEAMTLPKDAHIPVVEEHVMGKVLEEEKEAINQETVQQRDTFTPSGQEPILTEKETELKLEEKTTISDKEAVPKESKPPKPADEEIGIIQTSTEHTFSEQKDQEPTTDMLKQDSFPVSLEQAVTDSAMTSKTLEKAMTEPSALIEKSSIQELFEMRVDDKDKIEGVGAATSAELDMPFYEDKSGMSKYFETSALKEEATKSIEPGSDYYELSDTREBærerfriNFormuleringSupplied in 6 M Urea.Molekylvægt50 kDa (75 kDa by SDS-PAGE)Shipping temperatureShipped on blue ice at 4 °C.Tested applicationsSDS-PAGE, ELISA, Immunogen Grade - Specifikationer:
More
Specifications
Cat. No.ANTIA270588-50Protein/peptide nameMAP2 Projection P1Protein/peptide typeRecombinantSpeciesHumanKonjugationUnconjugatedGen IDUniprotID# P11137SekvensDLLTASKMEFHDQQELTPSTAEPSDQKEKESEKQSKPGEDLKHAALVSQPETTKTYPDKKDMQGTEEEKAPLALFGHTLVASLEDMKQKTEPSLVVPGIDLPKEPPTPKEQKDWFIEMPTEAKKDEWGLVAPISPGPLTPMREKDVFDDIPKWEGKQFDSPMPSPFQGGSFTLPLDVMKNEIVTETSPFAPAFLQPDDKKSLQQTSGPATAKDSFKIEEPHEAKPDKMAEAPPSEAMTLPKDAHIPVVEEHVMGKVLEEEKEAINQETVQQRDTFTPSGQEPILTEKETELKLEEKTTISDKEAVPKESKPPKPADEEIGIIQTSTEHTFSEQKDQEPTTDMLKQDSFPVSLEQAVTDSAMTSKTLEKAMTEPSALIEKSSIQELFEMRVDDKDKIEGVGAATSAELDMPFYEDKSGMSKYFETSALKEEATKSIEPGSDYYELSDTREBærerfriNFormuleringSupplied in 6 M Urea.Molekylvægt50 kDa (75 kDa by SDS-PAGE)Shipping temperatureShipped on blue ice at 4 °C.Tested applicationsSDS-PAGE, ELISA, Immunogen Grade
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



