Recombinant Human MAP2 Projection P2
Leverandør: Antibodies.com
Om denne vare
2 tilgængelige muligheder nedenfor
Product Details & Documents
Specifications for Product Family
Specifikationer
Specifikationer
- Catalog No:
- ANTIA270589-25
- ANTIA270589-50
- Protein/peptide name:
- MAP2 Projection P2
- MAP2 Projection P2
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Konjugation:
- Unconjugated
- Unconjugated
- Gen ID:
- UniprotID# P11137
- UniprotID# P11137
- Sekvens:
- SVHESIDTMSPMHKNGDKEFQTGKESQPSPPAQEAGYSTLAQSYPSDLPEEPSSPQERMFTIDPKVYGEKRDLHSKNKDDLTLSRSLGLGGRSAIEQRSMSINLPMSCLDSIALGFNFGRGHDLSPLASDILTNTSGSMDEGDDYLPATTPALEKAPCFPVESKEEEQIEKVKATGEESTQAEISCESPFLAKDFYKNGTVMAPDLPEMLDLAGTRSRLASVSADAEVARRKSVPSETVVEDSRTGLPPVTDENHVIVKTDSQLEDLGYCVFNKYTVPLPSPVQDSENLSGESGTFYEGTDDKVRRDLATDLSLIEVKLAAAGRVKDEFSVDKEASAHISGDKSGLSKEFDQEKKANDRLDTVLEKSEEHADSKEHAKKTEEAGDEIETFGLGVTYEQALAKDLSIPTDASSEKAEKGLSSVPEIAEVEPSKKVEQGLDFAVQGQLDVKISDFGQMASGLNIDDRRATELKLEATQDMTP
- SVHESIDTMSPMHKNGDKEFQTGKESQPSPPAQEAGYSTLAQSYPSDLPEEPSSPQERMFTIDPKVYGEKRDLHSKNKDDLTLSRSLGLGGRSAIEQRSMSINLPMSCLDSIALGFNFGRGHDLSPLASDILTNTSGSMDEGDDYLPATTPALEKAPCFPVESKEEEQIEKVKATGEESTQAEISCESPFLAKDFYKNGTVMAPDLPEMLDLAGTRSRLASVSADAEVARRKSVPSETVVEDSRTGLPPVTDENHVIVKTDSQLEDLGYCVFNKYTVPLPSPVQDSENLSGESGTFYEGTDDKVRRDLATDLSLIEVKLAAAGRVKDEFSVDKEASAHISGDKSGLSKEFDQEKKANDRLDTVLEKSEEHADSKEHAKKTEEAGDEIETFGLGVTYEQALAKDLSIPTDASSEKAEKGLSSVPEIAEVEPSKKVEQGLDFAVQGQLDVKISDFGQMASGLNIDDRRATELKLEATQDMTP
- Bærerfri:
- N
- N
- Shipping temperature:
- Shipped on blue ice at 4 °C.
- Shipped on blue ice at 4 °C.
- Tested applications:
- SDS-PAGE, ELISA
- SDS-PAGE, ELISA
Specifikationer
Product Family Options
Produktinformation
- PakningTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.ANTIA270589-25Protein/peptide nameMAP2 Projection P2Protein/peptide typeRecombinantSpeciesHumanKonjugationUnconjugatedGen IDUniprotID# P11137SekvensSVHESIDTMSPMHKNGDKEFQTGKESQPSPPAQEAGYSTLAQSYPSDLPEEPSSPQERMFTIDPKVYGEKRDLHSKNKDDLTLSRSLGLGGRSAIEQRSMSINLPMSCLDSIALGFNFGRGHDLSPLASDILTNTSGSMDEGDDYLPATTPALEKAPCFPVESKEEEQIEKVKATGEESTQAEISCESPFLAKDFYKNGTVMAPDLPEMLDLAGTRSRLASVSADAEVARRKSVPSETVVEDSRTGLPPVTDENHVIVKTDSQLEDLGYCVFNKYTVPLPSPVQDSENLSGESGTFYEGTDDKVRRDLATDLSLIEVKLAAAGRVKDEFSVDKEASAHISGDKSGLSKEFDQEKKANDRLDTVLEKSEEHADSKEHAKKTEEAGDEIETFGLGVTYEQALAKDLSIPTDASSEKAEKGLSSVPEIAEVEPSKKVEQGLDFAVQGQLDVKISDFGQMASGLNIDDRRATELKLEATQDMTPBærerfriNShipping temperatureShipped on blue ice at 4 °C.Tested applicationsSDS-PAGE, ELISA - Specifikationer:
More
Specifications
Cat. No.ANTIA270589-50Protein/peptide nameMAP2 Projection P2Protein/peptide typeRecombinantSpeciesHumanKonjugationUnconjugatedGen IDUniprotID# P11137SekvensSVHESIDTMSPMHKNGDKEFQTGKESQPSPPAQEAGYSTLAQSYPSDLPEEPSSPQERMFTIDPKVYGEKRDLHSKNKDDLTLSRSLGLGGRSAIEQRSMSINLPMSCLDSIALGFNFGRGHDLSPLASDILTNTSGSMDEGDDYLPATTPALEKAPCFPVESKEEEQIEKVKATGEESTQAEISCESPFLAKDFYKNGTVMAPDLPEMLDLAGTRSRLASVSADAEVARRKSVPSETVVEDSRTGLPPVTDENHVIVKTDSQLEDLGYCVFNKYTVPLPSPVQDSENLSGESGTFYEGTDDKVRRDLATDLSLIEVKLAAAGRVKDEFSVDKEASAHISGDKSGLSKEFDQEKKANDRLDTVLEKSEEHADSKEHAKKTEEAGDEIETFGLGVTYEQALAKDLSIPTDASSEKAEKGLSSVPEIAEVEPSKKVEQGLDFAVQGQLDVKISDFGQMASGLNIDDRRATELKLEATQDMTPBærerfriNShipping temperatureShipped on blue ice at 4 °C.Tested applicationsSDS-PAGE, ELISA
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



