Orders LinkContactUsLinkComponent
Adapter, delivery, bent, Quickfit®
Adapter, delivery, bent, Quickfit®
19/26 NS, 19/26 NS
Katalog # BIBBRA12/2
Leverandør:   SciLabware
Adapter, delivery, bent, Quickfit®
Katalog # BIBBRA12/2
Leverandør:  SciLabware
Adapter, delivery, bent, Quickfit®
Adapter, delivery, bent, Quickfit®
19/26 NS, 19/26 NS
Price
3.260,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
PYREX®, borosilicate glass, bend.
  • Excellent chemical resistance
  • High temperature resistance
Receiver RA12/2 is designed to accompany the vacuum receiver (RA11/SC) for four flasks shown in the image. It has an extension below the cone which is bent to direct the flow of condensate to the individual outlet chosen.
Specifikationer
  • Konusstr.:
    19/26 NS
  • Kappestr.:
    19/26 NS