Solid phase extraction cartridges, Bond Elut™ Plexa PCX
Om denne vare
Bond Elut™ Plexa PCX uses a polymeric cation exchange resin that combines the outstanding properties of Bond Elut™ Plexa - superior flow characteristics and improved analytical performance - with strong cation exchange functionalities. This mixed-mode SPE sorbent removes neutral and acidic interferences from the matrix, concentrates basic analytes and therefore improves sensitivity in the determination of basic compounds.
- Faster flow rates improve productivity
- Extraction cleanliness and reduced interference improve precision
- Simplified, single method
11 tilgængelige muligheder nedenfor
Product Details & Documents
Bond Elut™ Plexa PCX uses a polymeric cation exchange resin that combines the outstanding properties of Bond Elut™ Plexa - superior flow characteristics and improved analytical performance - with strong cation exchange functionalities. This mixed-mode SPE sorbent removes neutral and acidic interferences from the matrix, concentrates basic analytes and therefore improves sensitivity in the determination of basic compounds.
- Faster flow rates improve productivity
- Extraction cleanliness and reduced interference improve precision
- Simplified, single method
Specifications for Product Family
Specifikationer
- Catalog No:
- VARI12108301B
- VARI12108301
- 549-0808
- VARI12108303B
- VARI12108303
- 549-0809
- 549-0810
- VARI12108206
- 549-0838
- VARI12281306
- VARI5982-0603
- Surface area:
- 550 m²/g
- 550 m²/g
- 550 m²/g
- 550 m²/g
- 550 m²/g
- 550 m²/g
- 550 m²/g
- 550 m²/g
- 550 m²/g
- 550 m²/g
- 550 m²/g
Specifikationer
Product Family Options
Straight barrel cartridges
Produktinformation
- FasePorestørrelsePartikelstørrelseKapacitetLejevægtTilgængelighedPrice
- VARI12108301BStraight barrel cartridgeStraight barrel cartridgePlexa PCX100 Å45 µm1 ml30 mgLog ind for tilgængelighed1 * 1.000 Items29.420,00 krSpecifikationer:
More
Specifications
Cat. No.VARI12108301BSurface area550 m²/g - VARI12108301Straight barrel cartridgeStraight barrel cartridgePlexa PCX100 Å45 µm1 ml30 mgLog ind for tilgængelighed1 * 100 Items3.130,00 krSpecifikationer:
More
Specifications
Cat. No.VARI12108301Surface area550 m²/g - 549-0808Straight barrel cartridgeStraight barrel cartridgePlexa PCX100 Å45 µm1 ml60 mgLog ind for tilgængelighed1 * 100 Items3.180,00 krSpecifikationer:
More
Specifications
Cat. No.549-0808Surface area550 m²/g - VARI12108303BStraight barrel cartridgeStraight barrel cartridgePlexa PCX100 Å45 µm3 ml30 mgLog ind for tilgængelighed1 * 500 Items14.250,00 krSpecifikationer:
More
Specifications
Cat. No.VARI12108303BSurface area550 m²/g - VARI12108303Straight barrel cartridgeStraight barrel cartridgePlexa PCX100 Å45 µm3 ml30 mgLog ind for tilgængelighed1 * 50 Items1.620,00 krSpecifikationer:
More
Specifications
Cat. No.VARI12108303Surface area550 m²/g - 549-0809Straight barrel cartridgeStraight barrel cartridgePlexa PCX100 Å45 µm3 ml60 mgLog ind for tilgængelighed1 * 50 Items1.940,00 krSpecifikationer:
More
Specifications
Cat. No.549-0809Surface area550 m²/g - 549-0810Straight barrel cartridge, tablessStraight barrel cartridge, tablessPlexa PCX100 Å45 µm3 ml60 mgLog ind for tilgængelighed1 * 50 Items1.920,00 krSpecifikationer:
More
Specifications
Cat. No.549-0810Surface area550 m²/g - VARI12108206Straight barrel cartridgeStraight barrel cartridgePlexa PCX100 Å45 µm6 ml200 mgLog ind for tilgængelighed1 * 30 Items2.580,00 krSpecifikationer:
More
Specifications
Cat. No.VARI12108206Surface area550 m²/g - 549-0838Straight barrel cartridgeStraight barrel cartridgePlexa PCX100 Å45 µm6 ml500 mgLog ind for tilgængelighed1 * 30 Items3.840,00 krSpecifikationer:
More
Specifications
Cat. No.549-0838Surface area550 m²/g
Other formats
Produktinformation
- FasePorestørrelsePartikelstørrelseKapacitetLejevægtTilgængelighedPrice
- VARI12281306800 series cartridgeSpecifikationer:
More
Specifications
Cat. No.VARI12281306Surface area550 m²/g - VARI5982-0603LRC cartridgeSpecifikationer:
More
Specifications
Cat. No.VARI5982-0603Surface area550 m²/g
Bond Elut™ 96 round well plates
Produktinformation
- Katalog #BeskrivelsePåfyldningTilgængelighedPrice
- 96-brønds-plade, runde brønde, til SPE, Plexa™ PCX30 mgLog ind for tilgængelighed1 * 1 Items4.560,00 kr
Bond Elut™ 96 square well plates
Produktinformation
- Katalog #PåfyldningBeskrivelseTilgængelighedPrice
- 10 mg96-brønds-plade, firkantede brønde, til SPE, 2 ml, Plexa™ PCXLog ind for tilgængelighed1 * 1 Items4.660,00 kr