Thermal transfer printer labels, Type B-7423
Printer labels, B-7423, Polyester, Permanent akryl, Thermal transfer, hvid, 360X-Plus II, 5×5 mm, 5000 labels/roll
Katalog # BRDY622110
Leverandør: Brady
Gå til:
Product Details & Documents
Printable, polyester labels for thermal transfer printers.
Specifikationer
- Printer:360X-Plus II
- Printing method:Thermal transfer
- Model:B-7423
- Farve:hvid
- Beskrivelse:Printer labels
- Dimensioner:5×5 mm
- Materiale:Polyester
- Pakket:5000 labels/roll
- Klæbestof:Permanent akryl