Centrifuge tubes, pear shape, ASTM, PYREX®
Katalog # BIBB3440/02M
Leverandør: SciLabware
Specifikationer
- Lukkestørrelse:uden cap
- Ø×L:17×157 mm
- Capacity:100 ml
- Environmentally Preferable:
- Pakn:2
Specifikationer
Om denne vare
Centrifuge tube is used for the determination of water and sediment in petroleum products.
- Graduated in white enamel
Lower stem graduated from 0 to 1,5 ml in 0,1 ml divisions whereas upper body graduated from 1,5 to 5 ml in 0,5 ml divisions and from 5 to 10 ml in 1 ml.
Certifications: Complies with ASTM D96 and D-1966.