Orders LinkContactUsLinkComponent
Microtubes
Marketsource
Leverandør:   SARSTEDT
Microtubes
Leverandør:  SARSTEDT
Marketsource
Certifikater
12 tilgængelige muligheder nedenfor
Product Details & Documents
PP, translucent
  • Excellent chemical resistance
  • Excellent temperature resistance
Without or with skirted base.
Product Family Options
Tubes, 0,5 ml, type A, with skirted base, neutral loop cap attached
Produktinformation
  • Kapacitet
    Pakket
    Anbefalet arbejdsvolumen
    Tilgængelighed
    Price
  • SARS72.730.100
    Tubes with screw cap non assembled
    Tubes with screw cap non assembled
    0,5 ml
    5000/case
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 5.000 Items
    1 * 5.000 Items - 2.797,38 kr
    2.797,38 kr
    1 * 5.000 Items
       2.797,38 kr
    Specifikationer:
    Marketsource
    Marketsource
  • SARS72.730.106
    Tubes with screw cap assembled, sterile, with writing space
    Tubes with screw cap assembled, sterile, with writing space
    0,5 ml
    1000/case
    0,5 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items
    Specifikationer:
    Marketsource
    Marketsource
  • SARS72.730.105
    Tubes with screw cap assembled, sterile
    Tubes with screw cap assembled, sterile
    0,5 ml
    1000/case
    0,5 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items
    Specifikationer:
    Marketsource
    Marketsource
  • SARS72.730.107
    Tubes with screw cap non assembled, with writing space
    Tubes with screw cap non assembled, with writing space
    0,5 ml
    5000/case
    0,5 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 5.000 Items
    1 * 5.000 Items
    Specifikationer:
    Marketsource
    Marketsource
Tubes, 1,5 ml, type D, without skirted base, neutral loop cap attached
Produktinformation
  • Kapacitet
    Pakket
    Anbefalet arbejdsvolumen
    Tilgængelighed
    Price
  • SARS72.692.105
    Tubes with screw cap assembled, sterile
    Tubes with screw cap assembled, sterile
    1,5 ml
    1000/case
    1,5 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 1.166,09 kr
    1.166,09 kr
    1 * 1.000 Items
       1.166,09 kr
    Specifikationer:
    Marketsource
    Marketsource
  • SARS72.692.100
    Tubes with screw cap non assembled
    Tubes with screw cap non assembled
    1,5 ml
    5000/case
    1,5 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 5.000 Items
    1 * 5.000 Items
    Specifikationer:
    Marketsource
    Marketsource
Tubes, 2,0 ml, type H, without skirted base, neutral loop cap attached
Produktinformation
  • Kapacitet
    Pakket
    Anbefalet arbejdsvolumen
    Tilgængelighed
    Price
  • SARS72.693.105
    Tubes with screw cap assembled, sterile
    Tubes with screw cap assembled, sterile
    2,0 ml
    1000/case
    2,0 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items
    Specifikationer:
    Marketsource
    Marketsource
  • SARS72.693.100
    Tubes with screw cap non assembled
    Tubes with screw cap non assembled
    2,0 ml
    5000/case
    2,0 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 5.000 Items
    1 * 5.000 Items
    Specifikationer:
    Marketsource
    Marketsource
Tubes, 2,0 ml, type I, with skirted base, neutral loop cap attached
Produktinformation
  • Kapacitet
    Pakket
    Anbefalet arbejdsvolumen
    Tilgængelighed
    Price
  • SARS72.694.100
    Tubes with screw cap non assembled
    Tubes with screw cap non assembled
    2,0 ml
    5000/case
    2,0 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 5.000 Items
    1 * 5.000 Items
    Specifikationer:
    Marketsource
    Marketsource
  • SARS72.694.106
    Tubes with screw cap non assembled, with writing space and graduation
    Tubes with screw cap non assembled, with writing space and graduation
    2,0 ml
    1000/case
    2,0 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items
    Specifikationer:
    Marketsource
    Marketsource
  • SARS72.694.105
    Tubes with screw cap assembled, sterile
    Tubes with screw cap assembled, sterile
    2,0 ml
    1000/case
    2,0 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items
    Specifikationer:
    Marketsource
    Marketsource
  • SARS72.694.107
    Tubes with screw cap assembled, sterile, with writing space and graduation
    Tubes with screw cap assembled, sterile, with writing space and graduation
    2,0 ml
    5000/case
    2,0 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 5.000 Items
    1 * 5.000 Items
    Specifikationer:
    Marketsource
    Marketsource
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Colour coded inserts, blue
    Colour coded inserts, blue
    SARS65.713.006
    Log ind for tilgængelighed
    1 * 1.000 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Screw cap with seperate retaining loop, neutral
    Screw cap with seperate retaining loop, neutral
    SARS65.712
    Log ind for tilgængelighed
    1 * 5.000 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Colour coded inserts, neutral
    Colour coded inserts, neutral
    SARS65.713
    Log ind for tilgængelighed
    1 * 5.000 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Colour coded inserts, red
    Colour coded inserts, red
    SARS65.713.002
    Log ind for tilgængelighed
    1 * 5.000 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Colour coded inserts, violet
    Colour coded inserts, violet
    SARS65.713.007
    Log ind for tilgængelighed
    1 * 1.000 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Colour coded inserts, white
    Colour coded inserts, white
    SARS65.713.009
    Log ind for tilgængelighed
    1 * 1.000 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Colour coded inserts, black
    Colour coded inserts, black
    SARS65.713.010
    Log ind for tilgængelighed
    1 * 5.000 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Colour coded inserts, yellow
    Colour coded inserts, yellow
    SARS65.713.004
    Log ind for tilgængelighed
    1 * 5.000 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Colour coded inserts, green
    Colour coded inserts, green
    SARS65.713.005
    Log ind for tilgængelighed
    1 * 5.000 Items