Orders LinkContactUsLinkComponent
Adapters, pig receiver, Quickfit®
Adapters, pig receiver, Quickfit®
14/23 NS, 14/23 NS
Katalog # 271-0013
Leverandør:   SciLabware
Adapters, pig receiver, Quickfit®
Katalog # 271-0013
Leverandør:  SciLabware
Adapters, pig receiver, Quickfit®
Adapters, pig receiver, Quickfit®
14/23 NS, 14/23 NS
Price
1.500,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PYREX®, borosilicate glass
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Allows up to three flasks to be connected to a condenser outlet. Useful for collecting distillate fractions. By slightly rotating the adapter a new flask is presented to the distillate. With side-arm for connection to a vacuum supply.
Specifikationer
  • Konusstr.:
    14/23 NS
  • Kappestr.:
    14/23 NS