Orders LinkContactUsLinkComponent
Molecular model: ten-layer DNA, Molymod®
Molecular model: ten-layer DNA, Molymod®
Molecular model of DNA, 640
Catalog # SPIRMKS-122-10
Marketsource
Supplier:   SPIRING ENTERPRISES
Molecular model: ten-layer DNA, Molymod®
Catalog # SPIRMKS-122-10
Marketsource
Molecular model: ten-layer DNA, Molymod®
Molecular model: ten-layer DNA, Molymod®
Molecular model of DNA, 640
Price
1.785,84 kr
Log in for availability
1 * 1 Each
Product Details & Documents
Molecular model of compact DNA including ten pairs of bases.
  • Height ca. 50 cm
Ordering information: Self-assembly kit incl. 640 atoms and 750 connections on a stand.
Specifications
  • Beskrivelse:
    Molecular model of DNA
  • Antal atomer:
    640