Orders LinkContactUsLinkComponent
Autoclavable Bags
Autoclavable Bags
Autoclavable bags, PP, 660×307 mm
Katalog # 129-0006
Leverandør:   STERILIN
Autoclavable Bags
Katalog # 129-0006
Leverandør:  STERILIN
Autoclavable Bags
Autoclavable Bags
Autoclavable bags, PP, 660×307 mm
Price
1.450,00 kr
Log ind for tilgængelighed
1 * 200 Items
Product Details & Documents
The bags in PP can withstand temperatures up to 134 °C.
Specifikationer
  • Beskrivelse:
    Autoclavable bags
  • L×B:
    660×307 mm
  • Materiale:
    PP