Methylcyclohexan ≥99.0% (by GC) for HPLC
Glass bottle, 500 mL
Leverandør: TCI
Methylcyclohexan ≥99.0% (by GC) for HPLC
Katalog # TCIAM0627-500ML
Leverandør: TCI
CAS nummer: 108-87-2
Gå til:
Product Details & Documents
- CAS nummer:108-87-2
- Formel:C₆H₁₁CH₃
- MDL nummer:MFCD00001497
- Molecular Weight:98.19 g/mol
- Merck Index:14,06047
- Kogende Pt:100°C
- Smeltende Pt:–127°C
- Smeltende Pt:-127°C
- Tæthed:0,7693 g/cm³ (20 °C)
- Flash Pt:–4°C
- Flash Pt:-4°C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-624-3
- FN:2296
- ADR:3,II
Specifikationer
Specifikationstestresultater
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
- Absorbance (210nm):max. 1.000
- Absorbance (220nm):max. 0.500
- Absorbance (230nm):max. 0.200
Specifikationer
- Pakning:Glass bottle
- Pakn:500 mL
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



