Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Benzen ≥99.5% (by GC) sequencing solvent

Benzen ≥99.5% (by GC) sequencing solvent

Glass bottle, 500 mL
Katalog # TCIAQ0038-500ML
Brandfarlig
Irriterende
Sundhed
Miljø
Leverandør:   TCI
Benzen ≥99.5% (by GC) sequencing solvent
Katalog # TCIAQ0038-500ML
Leverandør:  TCI
CAS nummer:  71-43-2
Benzen ≥99.5% (by GC) sequencing solvent
Benzen ≥99.5% (by GC) sequencing solvent

Glass bottle, 500 mL

CAS nummer:  71-43-2
Price
206,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
Synonymer:  Cyclohexatriene
  • CAS nummer:71-43-2
  • Formel:C₆H₆
  • MDL nummer:MFCD00003009
  • Molecular Weight:78.11 g/mol
  • Merck Index:14,01066
  • Kogende Pt:80°C
  • Smeltende Pt:5,5 °C
  • Tæthed:0,878 g/cm³ (20 °C)
  • Flash Pt:–10°C
  • Flash Pt:-10°C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-753-7
  • FN:1114
  • ADR:3,II
  • NÅ:01-2119447106-44

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.5 %

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    500 mL

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.