Orders LinkContactUsLinkComponent
Inlet septa
Inlet septa
Inlet septa
Inlet septa
Inlet septa
These septa are available for a variety of applications and have different upper temperature limits. The lower temperature septa are usually softer, seal better and can withstand more punctures (injections) than their high temperature counterparts.
  • Made up low bleed silicone rubber formulations
  • Efficient by using sharp syringe needles
  • Septa should be replaced to avoid leaks and sample loss
  • To be used with recommended temperature range
Product Family Options
Non stick bleed and temperature optimized (BTO) septa
  • Beskrivelse
    Ø
  • Septa, bleed and temperature optimised (BTO), non-stick
    11 mm
    1 * 50 Items
    1 * 50 Items - 896,00 kr
    896,00 kr
    1 * 50 Items
       896,00 kr
  • Septa, bleed and temperature optimised (BTO), non-stick
    5 mm
    1 * 50 Items
    1 * 50 Items - 896,00 kr
    896,00 kr
    1 * 50 Items
       896,00 kr
  • Septa, bleed and temperature optimised (BTO), non-stick
    11 mm
    1 * 100 Items
    1 * 100 Items - 1.650,00 kr
    1.650,00 kr
    1 * 100 Items
       1.650,00 kr
This product has been blocked by your organization. Please contact your purchasing department for more information
Non stick advanced green septa
  • Beskrivelse
    Ø
  • Septa, Advanced Green, non-stick
    11 mm
    1 * 50 Items
    1 * 50 Items - 829,00 kr
    829,00 kr
    1 * 50 Items
       829,00 kr
  • Septa, Advanced Green, non-stick
    5 mm
    1 * 50 Items
    1 * 50 Items - 829,00 kr
    829,00 kr
    1 * 50 Items
       829,00 kr
  • Septa, Advanced Green, non-stick
    11 mm
    1 * 100 Items
    1 * 100 Items - 1.470,00 kr
    1.470,00 kr
    1 * 100 Items
       1.470,00 kr
This product has been blocked by your organization. Please contact your purchasing department for more information
Non stick long life septa
  • Beskrivelse
    Ø
  • Septa, long-life, non-stick
    11 mm
    1 * 50 Items
    1 * 50 Items - 1.010,00 kr
    1.010,00 kr
    1 * 50 Items
       1.010,00 kr
  • Septa, long-life, non-stick
    5 mm
    1 * 50 Items
    1 * 50 Items - 1.010,00 kr
    1.010,00 kr
    1 * 50 Items
       1.010,00 kr
  • Septa, long-life, non-stick
    11 mm
    1 * 100 Items
    1 * 100 Items - 1.800,00 kr
    1.800,00 kr
    1 * 100 Items
       1.800,00 kr
This product has been blocked by your organization. Please contact your purchasing department for more information
General purpose septa
  • Beskrivelse
    Ø
  • Septa, general purpose, grey
    5 mm
    1 * 25 Items
    1 * 25 Items - 351,00 kr
    351,00 kr
    1 * 25 Items
       351,00 kr
  • Septa, general purpose, grey
    5 mm
    1 * 25 Items
    1 * 25 Items - 344,00 kr
    344,00 kr
    1 * 25 Items
       344,00 kr
  • Septa, general purpose, red
    9,5 mm
    1 * 100 Items
    1 * 100 Items
  • Septa, general purpose, red
    9,5 mm
    1 * 50 Items
    1 * 50 Items - 627,00 kr
    627,00 kr
    1 * 50 Items
       627,00 kr
  • Septa, general purpose, grey
    11 mm
    1 * 100 Items
    1 * 100 Items - 1.130,00 kr
    1.130,00 kr
    1 * 100 Items
       1.130,00 kr
  • Septa, general purpose, grey
    9,5 mm
    1 * 100 Items
    1 * 100 Items - 1.130,00 kr
    1.130,00 kr
    1 * 100 Items
       1.130,00 kr
  • Septa, general purpose, red
    11 mm
    1 * 50 Items
    1 * 50 Items - 627,00 kr
    627,00 kr
    1 * 50 Items
       627,00 kr
  • Septa, general purpose, grey
    11 mm
    1 * 50 Items
    1 * 50 Items - 627,00 kr
    627,00 kr
    1 * 50 Items
       627,00 kr
  • Septa, general purpose, red
    11 mm
    1 * 100 Items
    1 * 100 Items - 1.130,00 kr
    1.130,00 kr
    1 * 100 Items
       1.130,00 kr
  • Septa, general purpose, grey
    9,5 mm
    1 * 50 Items
    1 * 50 Items - 627,00 kr
    627,00 kr
    1 * 50 Items
       627,00 kr
This product has been blocked by your organization. Please contact your purchasing department for more information
Merlin microseal
  • Beskrivelse
    Ø
  • Microseal septum, stainless steel, rubber (30 psi)
    1 * 1 Items
    1 * 1 Items - 1.760,00 kr
    1.760,00 kr
    1 * 1 Items
       1.760,00 kr
  • Merlin microseal kit, original low pressure system (includes nut and septum)
    1 * 1 Items
    1 * 1 Items - 3.380,00 kr
    3.380,00 kr
    1 * 1 Items
       3.380,00 kr
  • Merlin microseal kit, original low pressure system (includes nut and 2 septa)
    1 * 1 Items
    1 * 1 Items - 4.410,00 kr
    4.410,00 kr
    1 * 1 Items
       4.410,00 kr
  • High sample volume septum kit (contains Merlin high pressure Microseal, six 23-gauge syringes, 500 vials and caps)
    1 * 1 Items
    1 * 1 Items - 7.270,00 kr
    7.270,00 kr
    1 * 1 Items
       7.270,00 kr
  • Microseal PTFE nut liners
    1 * 2 Items
    1 * 2 Items - 97,00 kr
    97,00 kr
    1 * 2 Items
       97,00 kr
  • High pressure Merlin microseal starter kit (includes microseal septum and nut)
    1 * 1 Items
    1 * 1 Items - 3.540,00 kr
    3.540,00 kr
    1 * 1 Items
       3.540,00 kr
  • Merlin microseal high pressure septum
    1 * 1 Items
    1 * 1 Items - 2.000,00 kr
    2.000,00 kr
    1 * 1 Items
       2.000,00 kr
  • Microseal high pressure nut
    1 * 1 Items
    1 * 1 Items - 1.840,00 kr
    1.840,00 kr
    1 * 1 Items
       1.840,00 kr
This product has been blocked by your organization. Please contact your purchasing department for more information