Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries for LC systems
Leverandør:   AGILENT HEWLETT PACKARD
Capillaries for LC systems
Certifikater
Capillaries for LC systems
Capillaries for LC systems
109 tilgængelige muligheder nedenfor
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Product Family Options
Stainless steel capillaries
Produktinformation
  • Tilgængelighed
    Price
  • HEWLG1312-87300
    Absorber capillary, 500 μl, pre-swaged, male to male, 1770 mm, I.Ø 0,17 mm
    Absorber capillary, 500 μl, pre-swaged, male to male, 1770 mm, I.Ø 0,17 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 881,00 kr
    881,00 kr
    1 * 1 Items
       881,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1312-87300
  • HEWLG1312-67500
    Calibration capillary assembly
    Calibration capillary assembly
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 4.150,00 kr
    4.150,00 kr
    1 * 1 Items
       4.150,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1312-67500
  • HEWL01018-67305
    Capillaries, 1 end pre-swaged, blue, stainless steel, 700 mm, I.Ø 0,25 mm, to connect pump to universal
    Capillaries, 1 end pre-swaged, blue, stainless steel, 700 mm, I.Ø 0,25 mm, to connect pump to universal
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.040,00 kr
    1.040,00 kr
    1 * 1 Items
       1.040,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL01018-67305
  • HEWL01090-87305
    Capillaries, 1 end pre-swaged, green, stainless steel, 130 mm, I.Ø 0,17 mm, to connect autosampler to column
    Capillaries, 1 end pre-swaged, green, stainless steel, 130 mm, I.Ø 0,17 mm, to connect autosampler to column
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 806,00 kr
    806,00 kr
    1 * 1 Items
       806,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL01090-87305
  • HEWLG1313-87305
    Capillaries, 1 end pre-swaged, green, stainless steel, 180 mm, I.Ø 0,17 mm, to connect autosampler to column
    Capillaries, 1 end pre-swaged, green, stainless steel, 180 mm, I.Ø 0,17 mm, to connect autosampler to column
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 806,00 kr
    806,00 kr
    1 * 1 Items
       806,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1313-87305
  • HEWL01090-87304
    Capillaries, 1 end pre-swaged, green, stainless steel, 280 mm, I.Ø 0,17 mm, to connect autosampler to column
    Capillaries, 1 end pre-swaged, green, stainless steel, 280 mm, I.Ø 0,17 mm, to connect autosampler to column
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.180,00 kr
    1.180,00 kr
    1 * 1 Items
       1.180,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL01090-87304
  • HEWL01048-87302
    Capillaries, 1 end pre-swaged, green, stainless steel, 800 mm, I.Ø 0,17 mm, to connect autosampler to column
    Capillaries, 1 end pre-swaged, green, stainless steel, 800 mm, I.Ø 0,17 mm, to connect autosampler to column
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.040,00 kr
    1.040,00 kr
    1 * 1 Items
       1.040,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL01048-87302
  • HEWLG1329-87300
    Capillaries, 1 end pre-swaged, green, stainless steel, 900 mm, I.Ø 0,17 mm, to connect pump to manual valve
    Capillaries, 1 end pre-swaged, green, stainless steel, 900 mm, I.Ø 0,17 mm, to connect pump to manual valve
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 776,00 kr
    776,00 kr
    1 * 1 Items
       776,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1329-87300
  • HEWLG1316-87305
    Capillaries, 1 end pre-swaged, male to male, 340 mm, I.Ø 0,12 mm, to connect WPS to micro valve
    Capillaries, 1 end pre-swaged, male to male, 340 mm, I.Ø 0,12 mm, to connect WPS to micro valve
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 881,00 kr
    881,00 kr
    1 * 1 Items
       881,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1316-87305
  • HEWLG1315-87307
    Capillaries, 1 end pre-swaged, male to male, 500 mm, I.Ø 0,12 mm, to connect cell out to MS
    Capillaries, 1 end pre-swaged, male to male, 500 mm, I.Ø 0,12 mm, to connect cell out to MS
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 784,00 kr
    784,00 kr
    1 * 1 Items
       784,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1315-87307
  • HEWLG1316-87309
    Capillaries, 1 end pre-swaged, male to male, 500 mm, I.Ø 0,12 mm, to connect TCC/VWD to MS
    Capillaries, 1 end pre-swaged, male to male, 500 mm, I.Ø 0,12 mm, to connect TCC/VWD to MS
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 746,00 kr
    746,00 kr
    1 * 1 Items
       746,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1316-87309
  • HEWL01090-87611
    Capillaries, 1 end pre-swaged, red, stainless steel, 105 mm, I.Ø 0,12 mm, to connect autosampler to column
    Capillaries, 1 end pre-swaged, red, stainless steel, 105 mm, I.Ø 0,12 mm, to connect autosampler to column
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.170,00 kr
    1.170,00 kr
    1 * 1 Items
       1.170,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL01090-87611
  • HEWLG1313-87304
    Capillaries, 1 end pre-swaged, red, stainless steel, 180 mm, I.Ø 0,12 mm, to connect autosampler to column
    Capillaries, 1 end pre-swaged, red, stainless steel, 180 mm, I.Ø 0,12 mm, to connect autosampler to column
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 791,00 kr
    791,00 kr
    1 * 1 Items
       791,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1313-87304
  • HEWLG1373-87300
    Capillaries, 1 end pre-swaged, red, stainless steel, 250 mm, I.Ø 0,12 mm
    Capillaries, 1 end pre-swaged, red, stainless steel, 250 mm, I.Ø 0,12 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 612,00 kr
    612,00 kr
    1 * 1 Items
       612,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1373-87300
  • HEWL01090-87610
    Capillaries, 1 end pre-swaged, red, stainless steel, 280 mm, I.Ø 0,12 mm, to connect autosampler to column
    Capillaries, 1 end pre-swaged, red, stainless steel, 280 mm, I.Ø 0,12 mm, to connect autosampler to column
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.170,00 kr
    1.170,00 kr
    1 * 1 Items
       1.170,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL01090-87610
  • HEWLG1316-87303
    Capillaries, 1 end pre-swaged, red, stainless steel, 70 mm, I.Ø 0,12 mm, to connect column compartment to column
    Capillaries, 1 end pre-swaged, red, stainless steel, 70 mm, I.Ø 0,12 mm, to connect column compartment to column
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 791,00 kr
    791,00 kr
    1 * 1 Items
       791,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1316-87303
  • HEWLG1312-67301
    Capillaries, A/A, stainless steel, to connect damper to purge valve
    Capillaries, A/A, stainless steel, to connect damper to purge valve
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 724,00 kr
    724,00 kr
    1 * 1 Items
       724,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1312-67301
  • HEWLG1375-87400
    Capillaries, A/A, stainless steel, to connect filter to EMPV
    Capillaries, A/A, stainless steel, to connect filter to EMPV
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 791,00 kr
    791,00 kr
    1 * 1 Items
       791,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1375-87400
  • HEWLG1377-87301
    Capillaries, blue, stainless steel, 120 mm, I.Ø 0,25 mm, to connect injector valve to waste
    Capillaries, blue, stainless steel, 120 mm, I.Ø 0,25 mm, to connect injector valve to waste
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 717,00 kr
    717,00 kr
    1 * 1 Items
       717,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1377-87301
  • HEWLG1329-87101
    Capillaries, C, stainless steel, to connect needle seat to injection valve
    Capillaries, C, stainless steel, to connect needle seat to injection valve
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.590,00 kr
    2.590,00 kr
    1 * 1 Items
       2.590,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1329-87101
  • HEWLG1361-67306
    Capillaries, male to female, stainless steel, 0,6 mm, I.Ø 367 mm, to connect EMPV1 to union
    Capillaries, male to female, stainless steel, 0,6 mm, I.Ø 367 mm, to connect EMPV1 to union
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 843,00 kr
    843,00 kr
    1 * 1 Items
       843,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1361-67306
  • HEWLG1361-67304
    Capillaries, male to female, stainless steel, 111 mm, I.Ø 0,6 mm, to connect union to EMPV2
    Capillaries, male to female, stainless steel, 111 mm, I.Ø 0,6 mm, to connect union to EMPV2
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 843,00 kr
    843,00 kr
    1 * 1 Items
       843,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1361-67304
  • HEWLG1361-67305
    Capillaries, male to female, stainless steel, 40 mm, I.Ø 0,6 mm, to connect union to mixer
    Capillaries, male to female, stainless steel, 40 mm, I.Ø 0,6 mm, to connect union to mixer
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 843,00 kr
    843,00 kr
    1 * 1 Items
       843,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1361-67305
  • HEWLG1361-67300
    Capillaries, male to male, stainless steel, 173 mm, I.Ø 0,6 mm, to connect pump to 1 multi assembly
    Capillaries, male to male, stainless steel, 173 mm, I.Ø 0,6 mm, to connect pump to 1 multi assembly
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 843,00 kr
    843,00 kr
    1 * 1 Items
       843,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1361-67300
  • HEWLG1361-67301
    Capillaries, male to male, stainless steel, 175 mm, I.Ø 0,6 mm, to connect pump to 1 multi assembly
    Capillaries, male to male, stainless steel, 175 mm, I.Ø 0,6 mm, to connect pump to 1 multi assembly
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 843,00 kr
    843,00 kr
    1 * 1 Items
       843,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1361-67301
  • HEWLG1361-67302
    Capillaries, male to male, stainless steel, 400 mm, I.Ø 0,6 mm, to connect EMPV to next module
    Capillaries, male to male, stainless steel, 400 mm, I.Ø 0,6 mm, to connect EMPV to next module
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 843,00 kr
    843,00 kr
    1 * 1 Items
       843,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1361-67302
  • HEWLG1328-87600
    Capillaries, non-swaged, green, stainless steel, 500 mm, I.Ø 0,17 mm, to connect pump to autosampler
    Capillaries, non-swaged, green, stainless steel, 500 mm, I.Ø 0,17 mm, to connect pump to autosampler
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 791,00 kr
    791,00 kr
    1 * 1 Items
       791,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1328-87600
  • HEWLG1316-87300
    Capillaries, non-swaged, green, stainless steel, 90 mm, I.Ø 0,17 mm, to connect detector to waste
    Capillaries, non-swaged, green, stainless steel, 90 mm, I.Ø 0,17 mm, to connect detector to waste
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 627,00 kr
    627,00 kr
    1 * 1 Items
       627,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1316-87300
  • HEWLG1316-87315
    Capillaries, non-swaged, male to female, 130 mm, I.Ø 0,12 mm, to connect column to DAD cell
    Capillaries, non-swaged, male to female, 130 mm, I.Ø 0,12 mm, to connect column to DAD cell
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 717,00 kr
    717,00 kr
    1 * 1 Items
       717,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1316-87315
  • HEWLG1316-87304
    Capillaries, non-swaged, male to female, 130 mm, I.Ø 0,12 mm, to connect micro valve to regeneration pump
    Capillaries, non-swaged, male to female, 130 mm, I.Ø 0,12 mm, to connect micro valve to regeneration pump
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 881,00 kr
    881,00 kr
    1 * 1 Items
       881,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1316-87304
  • HEWLG1315-87303
    Capillaries, non-swaged, male to female, 150 mm, I.Ø 0,17 mm, to connect column to flow cell/DAD cell
    Capillaries, non-swaged, male to female, 150 mm, I.Ø 0,17 mm, to connect column to flow cell/DAD cell
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 717,00 kr
    717,00 kr
    1 * 1 Items
       717,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1315-87303
  • HEWLG1316-87327
    Capillaries, non-swaged, male to female, 170 mm, I.Ø 0,12 mm, to connect column to DAD cell
    Capillaries, non-swaged, male to female, 170 mm, I.Ø 0,12 mm, to connect column to DAD cell
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 717,00 kr
    717,00 kr
    1 * 1 Items
       717,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1316-87327
  • HEWLG1316-87312
    Capillaries, non-swaged, male to female, 50 mm, I.Ø 0,12 mm, to connect column to DAD cell
    Capillaries, non-swaged, male to female, 50 mm, I.Ø 0,12 mm, to connect column to DAD cell
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 717,00 kr
    717,00 kr
    1 * 1 Items
       717,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1316-87312
  • HEWLG1316-87313
    Capillaries, non-swaged, male to female, 70 mm, I.Ø 0,12 mm, to connect column to DAD cell
    Capillaries, non-swaged, male to female, 70 mm, I.Ø 0,12 mm, to connect column to DAD cell
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 717,00 kr
    717,00 kr
    1 * 1 Items
       717,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1316-87313
  • HEWLG1316-87326
    Capillaries, non-swaged, male to female, 75 mm, I.Ø 0,12 mm, to connect micro valve to detector
    Capillaries, non-swaged, male to female, 75 mm, I.Ø 0,12 mm, to connect micro valve to detector
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 746,00 kr
    746,00 kr
    1 * 1 Items
       746,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1316-87326
  • HEWLG1316-87314
    Capillaries, non-swaged, male to female, 90 mm, I.Ø 0,12 mm, to connect column to DAD cell
    Capillaries, non-swaged, male to female, 90 mm, I.Ø 0,12 mm, to connect column to DAD cell
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 717,00 kr
    717,00 kr
    1 * 1 Items
       717,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1316-87314
  • HEWLG1316-27301
    Capillaries, non-swaged, male to male, 100 mm, I.Ø 0,12 mm, to connect valve to valve
    Capillaries, non-swaged, male to male, 100 mm, I.Ø 0,12 mm, to connect valve to valve
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 784,00 kr
    784,00 kr
    1 * 1 Items
       784,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1316-27301
  • HEWLG1316-87321
    Capillaries, non-swaged, male to male, 105 mm, I.Ø 0,17 mm, to connect column to cooler (50-150 mm column)
    Capillaries, non-swaged, male to male, 105 mm, I.Ø 0,17 mm, to connect column to cooler (50-150 mm column)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 717,00 kr
    717,00 kr
    1 * 1 Items
       717,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1316-87321
  • HEWLG1316-87316
    Capillaries, non-swaged, male to male, 170 mm, I.Ø 0,12 mm, to connect WPS to TCC
    Capillaries, non-swaged, male to male, 170 mm, I.Ø 0,12 mm, to connect WPS to TCC
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 717,00 kr
    717,00 kr
    1 * 1 Items
       717,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1316-87316
  • HEWLG1316-87323
    Capillaries, non-swaged, male to male, 170 mm, I.Ø 0,17 mm, to connect column to cooler (20-30 mm column)
    Capillaries, non-swaged, male to male, 170 mm, I.Ø 0,17 mm, to connect column to cooler (20-30 mm column)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 717,00 kr
    717,00 kr
    1 * 1 Items
       717,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1316-87323
  • HEWLG1316-87317
    Capillaries, non-swaged, male to male, 210 mm, I.Ø 0,12 mm, to connect WPS to TCC
    Capillaries, non-swaged, male to male, 210 mm, I.Ø 0,12 mm, to connect WPS to TCC
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 717,00 kr
    717,00 kr
    1 * 1 Items
       717,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1316-87317
  • HEWLG1316-87318
    Capillaries, non-swaged, male to male, 300 mm, I.Ø 0,12 mm, to connect cooled WPS to TCC
    Capillaries, non-swaged, male to male, 300 mm, I.Ø 0,12 mm, to connect cooled WPS to TCC
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 717,00 kr
    717,00 kr
    1 * 1 Items
       717,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1316-87318
  • HEWLG1316-87319
    Capillaries, non-swaged, male to male, 340 mm, I.Ø 0,12 mm, to connect cooled WPS to TCC
    Capillaries, non-swaged, male to male, 340 mm, I.Ø 0,12 mm, to connect cooled WPS to TCC
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 717,00 kr
    717,00 kr
    1 * 1 Items
       717,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1316-87319
  • HEWLG1316-87306
    Capillaries, non-swaged, male to male, 75 mm, I.Ø 0,12 mm, to connect micro valve to heat exchanger
    Capillaries, non-swaged, male to male, 75 mm, I.Ø 0,12 mm, to connect micro valve to heat exchanger
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 746,00 kr
    746,00 kr
    1 * 1 Items
       746,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1316-87306
  • HEWL01090-87308
    Capillaries, pre-swaged, blue, stainless steel, 130 mm, I.Ø 0,25 mm, to connect pump to autosampler
    Capillaries, pre-swaged, blue, stainless steel, 130 mm, I.Ø 0,25 mm, to connect pump to autosampler
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.360,00 kr
    1.360,00 kr
    1 * 1 Items
       1.360,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL01090-87308
  • HEWL79835-87638
    Capillaries, pre-swaged, blue, stainless steel, 320 mm, I.Ø 0,25 mm, to connect pump to autosampler
    Capillaries, pre-swaged, blue, stainless steel, 320 mm, I.Ø 0,25 mm, to connect pump to autosampler
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.460,00 kr
    1.460,00 kr
    1 * 1 Items
       1.460,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL79835-87638
  • HEWLG1367-87304
    Capillaries, pre-swaged, green, stainless steel, 250 mm, I.Ø 0,17 mm
    Capillaries, pre-swaged, green, stainless steel, 250 mm, I.Ø 0,17 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 717,00 kr
    717,00 kr
    1 * 1 Items
       717,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1367-87304
  • HEWLG1315-87311
    Capillaries, pre-swaged, green, stainless steel, 380 mm, I.Ø 0,17 mm, to connect column to DAD
    Capillaries, pre-swaged, green, stainless steel, 380 mm, I.Ø 0,17 mm, to connect column to DAD
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.060,00 kr
    1.060,00 kr
    1 * 1 Items
       1.060,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1315-87311
  • HEWLG1312-67305
    Capillaries, pre-swaged, green, stainless steel, 600 mm, I.Ø 0,17 mm, to connect pump to autosampler
    Capillaries, pre-swaged, green, stainless steel, 600 mm, I.Ø 0,17 mm, to connect pump to autosampler
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 776,00 kr
    776,00 kr
    1 * 1 Items
       776,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1312-67305
  • HEWLG1312-87306
    Capillaries, pre-swaged, male to male, 105 mm, I.Ø 0,17 mm, to connect damper/mixer to mixer/outlet valve
    Capillaries, pre-swaged, male to male, 105 mm, I.Ø 0,17 mm, to connect damper/mixer to mixer/outlet valve
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 717,00 kr
    717,00 kr
    1 * 1 Items
       717,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1312-87306
  • HEWLG1312-87305
    Capillaries, pre-swaged, male to male, 150 mm, I.Ø 0,17 mm, to connect damper to pressure sensor
    Capillaries, pre-swaged, male to male, 150 mm, I.Ø 0,17 mm, to connect damper to pressure sensor
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 717,00 kr
    717,00 kr
    1 * 1 Items
       717,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1312-87305
  • HEWLG1367-87303
    Capillaries, pre-swaged, male to male, 210 mm, I.Ø 0,12 mm, to connect WPS to 10 port valve
    Capillaries, pre-swaged, male to male, 210 mm, I.Ø 0,12 mm, to connect WPS to 10 port valve
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 717,00 kr
    717,00 kr
    1 * 1 Items
       717,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1367-87303
  • HEWL5067-1571
    Capillaries, pre-swaged, male to male, 320 mm, I.Ø 0,17 mm
    Capillaries, pre-swaged, male to male, 320 mm, I.Ø 0,17 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 717,00 kr
    717,00 kr
    1 * 1 Items
       717,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5067-1571
  • HEWLG1312-87303
    Capillaries, pre-swaged, male to male, 400 mm, I.Ø 0,17 mm, to connect pump to autosampler
    Capillaries, pre-swaged, male to male, 400 mm, I.Ø 0,17 mm, to connect pump to autosampler
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 717,00 kr
    717,00 kr
    1 * 1 Items
       717,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1312-87303
  • HEWLG1312-87304
    Capillaries, pre-swaged, male to male, 700 mm, I.Ø 0,17 mm, to connect pump to cooled autosampler
    Capillaries, pre-swaged, male to male, 700 mm, I.Ø 0,17 mm, to connect pump to cooled autosampler
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 746,00 kr
    746,00 kr
    1 * 1 Items
       746,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1312-87304
  • HEWLG1311-67304
    Capillaries, pre-swaged, PTFE, to connect SSV/MCGV to AIV
    Capillaries, pre-swaged, PTFE, to connect SSV/MCGV to AIV
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 746,00 kr
    746,00 kr
    1 * 1 Items
       746,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1311-67304
  • HEWLG1315-87312
    Capillaries, pre-swaged, red, stainless steel, 150 mm, I.Ø 0,12 mm, to connect column to DAD
    Capillaries, pre-swaged, red, stainless steel, 150 mm, I.Ø 0,12 mm, to connect column to DAD
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.060,00 kr
    1.060,00 kr
    1 * 1 Items
       1.060,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1315-87312
  • HEWLG1316-87328
    Capillaries, pre-swaged, red, stainless steel, 210 mm, I.Ø 0,12 mm
    Capillaries, pre-swaged, red, stainless steel, 210 mm, I.Ø 0,12 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 724,00 kr
    724,00 kr
    1 * 1 Items
       724,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1316-87328
  • HEWLG2258-87301
    Capillaries, stainless steel, 160 mm, I.Ø 0,5 mm, to connect injector valve to prep head
    Capillaries, stainless steel, 160 mm, I.Ø 0,5 mm, to connect injector valve to prep head
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 776,00 kr
    776,00 kr
    1 * 1 Items
       776,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG2258-87301
  • HEWLG2260-87300
    Capillaries, stainless steel, 600 mm, I.Ø 0,5 mm, to connect autosampler to column
    Capillaries, stainless steel, 600 mm, I.Ø 0,5 mm, to connect autosampler to column
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 806,00 kr
    806,00 kr
    1 * 1 Items
       806,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG2260-87300
  • HEWL5062-8522
    Capillaries, with fittings, PEEK, 40 mm, I.Ø 0,18 mm, to connect column to VWD
    Capillaries, with fittings, PEEK, 40 mm, I.Ø 0,18 mm, to connect column to VWD
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 456,00 kr
    456,00 kr
    1 * 1 Items
       456,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5062-8522
  • HEWL5021-1816
    Capillaries, without fittings, green, stainless steel, 105 mm, I.Ø 0,17 mm, to connect column to VWD
    Capillaries, without fittings, green, stainless steel, 105 mm, I.Ø 0,17 mm, to connect column to VWD
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 351,00 kr
    351,00 kr
    1 * 1 Items
       351,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5021-1816
  • HEWL5021-1817
    Capillaries, without fittings, green, stainless steel, 150 mm, I.Ø 0,17 mm, to connect column to VWD
    Capillaries, without fittings, green, stainless steel, 150 mm, I.Ø 0,17 mm, to connect column to VWD
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 351,00 kr
    351,00 kr
    1 * 1 Items
       351,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5021-1817
  • HEWL5021-1818
    Capillaries, without fittings, green, stainless steel, 280 mm, I.Ø 0,17 mm, to connect column to VWD
    Capillaries, without fittings, green, stainless steel, 280 mm, I.Ø 0,17 mm, to connect column to VWD
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 351,00 kr
    351,00 kr
    1 * 1 Items
       351,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5021-1818
  • HEWL5021-1819
    Capillaries, without fittings, green, stainless steel, 400 mm, I.Ø 0,17 mm, to connect column to VWD
    Capillaries, without fittings, green, stainless steel, 400 mm, I.Ø 0,17 mm, to connect column to VWD
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 351,00 kr
    351,00 kr
    1 * 1 Items
       351,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5021-1819
  • HEWL5062-8535
    Capillaries, without fittings, PEEK, 48 mm, I.Ø 0,25 mm, to connect VWD to waste
    Capillaries, without fittings, PEEK, 48 mm, I.Ø 0,25 mm, to connect VWD to waste
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 709,00 kr
    709,00 kr
    1 * 1 Items
       709,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5062-8535
  • HEWL5021-1820
    Capillaries, without fittings, red, stainless steel, 105 mm, I.Ø 0,12 mm, to connect column to VWD
    Capillaries, without fittings, red, stainless steel, 105 mm, I.Ø 0,12 mm, to connect column to VWD
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 351,00 kr
    351,00 kr
    1 * 1 Items
       351,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5021-1820
  • HEWL5021-1821
    Capillaries, without fittings, red, stainless steel, 150 mm, I.Ø 0,12 mm, to connect column to VWD
    Capillaries, without fittings, red, stainless steel, 150 mm, I.Ø 0,12 mm, to connect column to VWD
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 351,00 kr
    351,00 kr
    1 * 1 Items
       351,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5021-1821
  • HEWL5021-1822
    Capillaries, without fittings, red, stainless steel, 280 mm, I.Ø 0,12 mm, to connect column to VWD
    Capillaries, without fittings, red, stainless steel, 280 mm, I.Ø 0,12 mm, to connect column to VWD
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 351,00 kr
    351,00 kr
    1 * 1 Items
       351,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5021-1822
  • HEWL5021-1823
    Capillaries, without fittings, red, stainless steel, 400 mm, I.Ø 0,12 mm, to connect column to VWD
    Capillaries, without fittings, red, stainless steel, 400 mm, I.Ø 0,12 mm, to connect column to VWD
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 351,00 kr
    351,00 kr
    1 * 1 Items
       351,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5021-1823
  • HEWL79826-87606
    Capillary, stainless steel, I.Ø 0,17 mm, 80 mm
    Capillary, stainless steel, I.Ø 0,17 mm, 80 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.200,00 kr
    1.200,00 kr
    1 * 1 Items
       1.200,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL79826-87606
  • HEWLG1315-87339
    DAD heat exchanger capillary, 1 end pre-swaged, male to male, 310 mm, I.Ø 0,12 mm
    DAD heat exchanger capillary, 1 end pre-swaged, male to male, 310 mm, I.Ø 0,12 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.000,00 kr
    1.000,00 kr
    1 * 1 Items
       1.000,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1315-87339
  • HEWLG1315-87319
    DAD heat exchanger capillary, 1 end pre-swaged, male to male, 310 mm, I.Ø 0,17 mm
    DAD heat exchanger capillary, 1 end pre-swaged, male to male, 310 mm, I.Ø 0,17 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.000,00 kr
    1.000,00 kr
    1 * 1 Items
       1.000,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1315-87319
  • HEWL5065-9979
    Flexible capillaries, blue, stainless steel, without fittings, 12,3 μl, 250 mm, I.Ø 0,25 mm
    Flexible capillaries, blue, stainless steel, without fittings, 12,3 μl, 250 mm, I.Ø 0,25 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 351,00 kr
    351,00 kr
    1 * 1 Items
       351,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5065-9979
  • HEWL5022-6508
    Flexible capillaries, blue, stainless steel, without fittings, 13,8 μl, 280 mm, I.Ø 0,25 mm
    Flexible capillaries, blue, stainless steel, without fittings, 13,8 μl, 280 mm, I.Ø 0,25 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 351,00 kr
    351,00 kr
    1 * 1 Items
       351,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5022-6508
  • HEWL5065-9980
    Flexible capillaries, blue, stainless steel, without fittings, 15,8 μl, 320 mm, I.Ø 0,25 mm
    Flexible capillaries, blue, stainless steel, without fittings, 15,8 μl, 320 mm, I.Ø 0,25 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 351,00 kr
    351,00 kr
    1 * 1 Items
       351,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5065-9980
  • HEWL5065-9930
    Flexible capillaries, blue, stainless steel, without fittings, 39,3 μl, 800 mm, I.Ø 0,25 mm
    Flexible capillaries, blue, stainless steel, without fittings, 39,3 μl, 800 mm, I.Ø 0,25 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 396,00 kr
    396,00 kr
    1 * 1 Items
       396,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5065-9930
  • HEWL5065-9933
    Flexible capillaries, green, stainless steel, without fittings, 13,6 μl, 600 mm, I.Ø 0,17 mm
    Flexible capillaries, green, stainless steel, without fittings, 13,6 μl, 600 mm, I.Ø 0,17 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 396,00 kr
    396,00 kr
    1 * 1 Items
       396,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5065-9933
  • HEWL5065-9932
    Flexible capillaries, green, stainless steel, without fittings, 15,9 μl, 700 mm, I.Ø 0,17 mm
    Flexible capillaries, green, stainless steel, without fittings, 15,9 μl, 700 mm, I.Ø 0,17 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 396,00 kr
    396,00 kr
    1 * 1 Items
       396,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5065-9932
  • HEWLG1315-87306
    Capillaries tubing, stainless steel, 200 mm, 0.12 mm Int.Ø
    Capillaries tubing, stainless steel, 200 mm, 0.12 mm Int.Ø
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.060,00 kr
    1.060,00 kr
    1 * 1 Items
       1.060,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1315-87306
  • HEWL5065-9963
    Flexible capillaries, green, stainless steel, without fittings, 20,5 μl, 900 mm, I.Ø 0,17 mm
    Flexible capillaries, green, stainless steel, without fittings, 20,5 μl, 900 mm, I.Ø 0,17 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 351,00 kr
    351,00 kr
    1 * 1 Items
       351,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5065-9963
  • HEWL5065-9931
    Flexible capillaries, green, stainless steel, without fittings, 4,6 μl, 200 mm, I.Ø 0,17 mm
    Flexible capillaries, green, stainless steel, without fittings, 4,6 μl, 200 mm, I.Ø 0,17 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 351,00 kr
    351,00 kr
    1 * 1 Items
       351,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5065-9931
  • HEWL5065-9935
    Flexible capillaries, red, stainless steel, without fittings, 2,3 μl, 200 mm, I.Ø 0,12 mm
    Flexible capillaries, red, stainless steel, without fittings, 2,3 μl, 200 mm, I.Ø 0,12 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 351,00 kr
    351,00 kr
    1 * 1 Items
       351,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5065-9935
  • HEWL5065-9964
    Flexible capillaries, red, stainless steel, without fittings, 5,6 μl, 500 mm, I.Ø 0,12 mm
    Flexible capillaries, red, stainless steel, without fittings, 5,6 μl, 500 mm, I.Ø 0,12 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 351,00 kr
    351,00 kr
    1 * 1 Items
       351,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5065-9964
  • HEWL5065-9926
    Flexible capillaries, stainless steel, without fittings, 157 μl, 800 mm, I.Ø 0,5 mm
    Flexible capillaries, stainless steel, without fittings, 157 μl, 800 mm, I.Ø 0,5 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 814,00 kr
    814,00 kr
    1 * 1 Items
       814,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5065-9926
  • HEWL5065-9927
    Flexible capillaries, stainless steel, without fittings, 20,6 μl, 105 mm, I.Ø 0,5 mm
    Flexible capillaries, stainless steel, without fittings, 20,6 μl, 105 mm, I.Ø 0,5 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 351,00 kr
    351,00 kr
    1 * 1 Items
       351,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5065-9927
  • HEWL5022-6509
    Flexible capillaries, stainless steel, without fittings, 29,5 μl, 150 mm, I.Ø 0,5 mm
    Flexible capillaries, stainless steel, without fittings, 29,5 μl, 150 mm, I.Ø 0,5 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 351,00 kr
    351,00 kr
    1 * 1 Items
       351,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5022-6509
  • HEWL5022-6510
    Flexible capillaries, stainless steel, without fittings, 55 μl, 280 mm, I.Ø 0,5 mm
    Flexible capillaries, stainless steel, without fittings, 55 μl, 280 mm, I.Ø 0,5 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 351,00 kr
    351,00 kr
    1 * 1 Items
       351,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5022-6510
  • HEWL5022-2159
    Restriction capillaries, stainless steel, with 1/16” female connector on both ends, 2000 mm, I.Ø 0,12 mm
    Restriction capillaries, stainless steel, with 1/16” female connector on both ends, 2000 mm, I.Ø 0,12 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.210,00 kr
    1.210,00 kr
    1 * 1 Items
       1.210,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWL5022-2159
  • HEWLG1312-87301
    Restriction capillary, I.Ø 0,17 mm
    Restriction capillary, I.Ø 0,17 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.280,00 kr
    1.280,00 kr
    1 * 1 Items
       1.280,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1312-87301
PEEK coated fused silica capillaries
Produktinformation
  • Tilgængelighed
    Price
  • HEWLG1315-68708
    Capillaries, PEEK coated fused silica, E/-, 700 mm, I.Ø 1905 mm, to connect detector to waste
    Capillaries, PEEK coated fused silica, E/-, 700 mm, I.Ø 1905 mm, to connect detector to waste
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 2 Items
    1 * 2 Items - 1.150,00 kr
    1.150,00 kr
    1 * 2 Items
       1.150,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1315-68708
  • HEWLG1375-87301
    Capillaries, PEEK coated fused silica, B/B, 220 mm, I.Ø 1270 mm, to connect EMPV to flow sensor
    Capillaries, PEEK coated fused silica, B/B, 220 mm, I.Ø 1270 mm, to connect EMPV to flow sensor
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.150,00 kr
    1.150,00 kr
    1 * 1 Items
       1.150,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1375-87301
  • HEWLG1375-87302
    Capillaries, PEEK coated fused silica, B/C, 200 mm, I.Ø 1270 mm, to connect injection valve to metering device
    Capillaries, PEEK coated fused silica, B/C, 200 mm, I.Ø 1270 mm, to connect injection valve to metering device
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 926,00 kr
    926,00 kr
    1 * 1 Items
       926,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1375-87302
  • HEWLG1375-87303
    Capillaries, PEEK coated fused silica, B/B, 110 mm, I.Ø 2540 mm, to connect metering device to needle
    Capillaries, PEEK coated fused silica, B/B, 110 mm, I.Ø 2540 mm, to connect metering device to needle
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.390,00 kr
    1.390,00 kr
    1 * 1 Items
       1.390,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1375-87303
  • HEWLG1375-87304
    Capillaries, PEEK coated fused silica, C/D, 500 mm, I.Ø 1270 mm, to connect injection valve to column
    Capillaries, PEEK coated fused silica, C/D, 500 mm, I.Ø 1270 mm, to connect injection valve to column
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.150,00 kr
    1.150,00 kr
    1 * 1 Items
       1.150,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1375-87304
  • HEWLG1375-87305
    Capillaries, PEEK coated fused silica, B/B, 220 mm, I.Ø 2540 mm, to connect EMPV to flow sensor
    Capillaries, PEEK coated fused silica, B/B, 220 mm, I.Ø 2540 mm, to connect EMPV to flow sensor
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.150,00 kr
    1.150,00 kr
    1 * 1 Items
       1.150,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1375-87305
  • HEWLG1375-87306
    Capillaries, PEEK coated fused silica, B/C, 550 mm, I.Ø 2540 mm, to connect flow sensor to injection valve
    Capillaries, PEEK coated fused silica, B/C, 550 mm, I.Ø 2540 mm, to connect flow sensor to injection valve
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.150,00 kr
    1.150,00 kr
    1 * 1 Items
       1.150,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1375-87306
  • HEWLG1375-87308
    Capillaries, PEEK coated fused silica, D/E, 400 mm, I.Ø 1905 mm, to connect column to waste
    Capillaries, PEEK coated fused silica, D/E, 400 mm, I.Ø 1905 mm, to connect column to waste
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.150,00 kr
    1.150,00 kr
    1 * 1 Items
       1.150,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1375-87308
  • HEWLG1375-87309
    Capillaries, PEEK coated fused silica, C/D, 280 mm, I.Ø 1270 mm, to connect μ-switching valve to column
    Capillaries, PEEK coated fused silica, C/D, 280 mm, I.Ø 1270 mm, to connect μ-switching valve to column
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 926,00 kr
    926,00 kr
    1 * 1 Items
       926,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1375-87309
  • HEWLG1375-87310
    Capillaries, PEEK coated fused silica, B/C, 550 mm, I.Ø 1270 mm, to connect flow sensor to injection valve
    Capillaries, PEEK coated fused silica, B/C, 550 mm, I.Ø 1270 mm, to connect flow sensor to injection valve
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.150,00 kr
    1.150,00 kr
    1 * 1 Items
       1.150,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1375-87310
  • HEWLG1375-87311
    Capillaries, PEEK coated fused silica, C/D, 500 mm, I.Ø 1905 mm, to connect injection valve to column
    Capillaries, PEEK coated fused silica, C/D, 500 mm, I.Ø 1905 mm, to connect injection valve to column
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.150,00 kr
    1.150,00 kr
    1 * 1 Items
       1.150,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1375-87311
  • HEWLG1375-87312
    Capillaries, PEEK coated fused silica, B/C, 200 mm, I.Ø 2540 mm, to connect injection valve to metering device
    Capillaries, PEEK coated fused silica, B/C, 200 mm, I.Ø 2540 mm, to connect injection valve to metering device
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 926,00 kr
    926,00 kr
    1 * 1 Items
       926,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1375-87312
  • HEWLG1375-87320
    Capillaries, PEEK coated fused silica, C/D, 100 mm, I.Ø 635 mm, to connect switching valve to column
    Capillaries, PEEK coated fused silica, C/D, 100 mm, I.Ø 635 mm, to connect switching valve to column
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 926,00 kr
    926,00 kr
    1 * 1 Items
       926,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1375-87320
  • HEWLG1375-87321
    Capillaries, PEEK coated fused silica, B/B, 220 mm, I.Ø 635 mm, to connect EMPV to flow sensor
    Capillaries, PEEK coated fused silica, B/B, 220 mm, I.Ø 635 mm, to connect EMPV to flow sensor
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.150,00 kr
    1.150,00 kr
    1 * 1 Items
       1.150,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1375-87321
  • HEWLG1375-87322
    Capillaries, PEEK coated fused silica, B/C, 350 mm, I.Ø 635 mm, to connect flow sensor to injection valve
    Capillaries, PEEK coated fused silica, B/C, 350 mm, I.Ø 635 mm, to connect flow sensor to injection valve
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.150,00 kr
    1.150,00 kr
    1 * 1 Items
       1.150,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1375-87322
  • HEWLG1375-87323
    Capillaries, PEEK coated fused silica, C/D, 550 mm, I.Ø 635 mm, to connect injection valve/switching valve to flow sensor/column
    Capillaries, PEEK coated fused silica, C/D, 550 mm, I.Ø 635 mm, to connect injection valve/switching valve to flow sensor/column
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.150,00 kr
    1.150,00 kr
    1 * 1 Items
       1.150,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1375-87323
  • HEWLG1375-87324
    Capillaries, PEEK coated fused silica, C/D, 700 mm, I.Ø 635 mm, to connect switching valve to column
    Capillaries, PEEK coated fused silica, C/D, 700 mm, I.Ø 635 mm, to connect switching valve to column
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.390,00 kr
    1.390,00 kr
    1 * 1 Items
       1.390,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1375-87324
  • HEWLG1375-87325
    Capillaries, PEEK coated fused silica, C/D, 100 mm, I.Ø 1270 mm, to connect switching valve to column
    Capillaries, PEEK coated fused silica, C/D, 100 mm, I.Ø 1270 mm, to connect switching valve to column
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 926,00 kr
    926,00 kr
    1 * 1 Items
       926,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1375-87325
  • HEWLG1375-87327
    Capillaries, PEEK coated fused silica, C/D, 650 mm, I.Ø 1905 mm, to connect injection valve to injector seat or 2nd pump
    Capillaries, PEEK coated fused silica, C/D, 650 mm, I.Ø 1905 mm, to connect injection valve to injector seat or 2nd pump
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.390,00 kr
    1.390,00 kr
    1 * 1 Items
       1.390,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    HEWLG1375-87327