Fittings
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Om denne vare
56 tilgængelige muligheder nedenfor
Product Details & Documents
Product Family Options
Nuts, front and back ferrule sets
Produktinformation
- TilgængelighedPrice
- HEWL5180-41491/16" stainless steel nuts, front and back ferrule sets1/16" stainless steel nuts, front and back ferrule setsLog ind for tilgængelighed1 * 10 Items3.700,00 kr
- HEWL5080-87501/8" brass nuts, front and back ferrule sets
- HEWL5080-87511/8" stainless steel nuts, front and back ferrule sets1/8" stainless steel nuts, front and back ferrule setsLog ind for tilgængelighed1 * 20 Items4.100,00 kr
- HEWL5080-87521/4" brass nuts, front and back ferrule sets
- HEWL5080-87531/4" stainless steel nuts, front and back ferrule sets1/4" stainless steel nuts, front and back ferrule setsLog ind for tilgængelighed1 * 20 Items3.700,00 kr
Nuts
Produktinformation
- TilgængelighedPrice
- HEWL0100-09291/16" stainless steel male nuts
- HEWL5180-41021/16" stainless steel male nuts
- HEWL5180-41031/8" brass nuts
- HEWL5180-41051/4" brass nuts
Front ferrules
Produktinformation
- TilgængelighedPrice
- HEWL5180-41081/16" stainless steel front ferrules
- HEWL5181-12921/16" stainless steel front ferrules, Valco one piece1/16" stainless steel front ferrules, Valco one pieceLog ind for tilgængelighed1 * 10 Items538,00 kr
- HEWL5180-41091/8" brass front ferrules
- HEWL5180-41111/4" brass front ferrules
Back ferrules
Produktinformation
- TilgængelighedPrice
- HEWL5180-41141/16" stainless steel back ferrules
- HEWL5180-41151/8" brass back ferrules
- HEWL5180-41171/4" brass back ferrules
Caps
Produktinformation
- TilgængelighedPrice
- HEWL5180-41201/4" brass caps
- HEWL5180-41211/8" brass caps
Plugs
Produktinformation
- TilgængelighedPrice
- HEWL5180-41241/8" brass plugs
- HEWL5180-41251/4" brass plugs
Blanking nut, adapter, hose connector
Produktinformation
- TilgængelighedPrice
- HEWL01080-832021/16" stainless steel blanking nut
- HEWL0100-04201/8"×1/8" brass adapter
- HEWL5180-41481/16" brass hose connectors
Tees
Produktinformation
- TilgængelighedPrice
- HEWL5180-41291/4" brass tees
- HEWL0100-00891/4" stainless steel tee
- HEWL5180-41601/8" brass tees
- HEWL0100-05421/8" stainless steel tee
- HEWL0100-06801/16" brass tees
- HEWL0100-07821/16" stainless steel tee
- HEWL0100-09691/16" stainless steel tee, low dead volume
Crosses
Produktinformation
- TilgængelighedPrice
- HEWL0100-01611/8" brass crosses
- HEWL0100-09851/4" brass crosses
Elbows
Produktinformation
- TilgængelighedPrice
- HEWL0100-0091¹/₈" brass elbow
Unions
Produktinformation
- TilgængelighedPrice
- HEWL0100-01241/16" stainless steel union
- HEWL0100-01261/8" stainless steel union
- HEWL0100-01281/4" stainless steel union
- HEWL0100-13161/16" brass union
- HEWL5180-41271/8" brass unions
- HEWL5180-41281/4" brass unions
Zero dead volume unions
Produktinformation
- TilgængelighedPrice
- HEWL0100-09001/16" stainless steel union, zero dead volume
- HEWL0100-1619Stainless steel union, zero dead volume"
Bulkhead unions
Produktinformation
- TilgængelighedPrice
- HEWL0100-01321/8" stainless steel bulkhead union
- HEWL0100-01331/8" brass bulkhead union
Connectors (Swagelok to male NPT)
Produktinformation
- TilgængelighedPrice
- HEWL0100-01111/8" (A)×1/8" (B) stainless steel connector
- HEWL0100-11891/4" (A)×1/4" (B) stainless steel connector
- HEWL5180-41431/8" (A)×1/4" (B) brass connector
- HEWL5180-41451/4" (A)×1/4" (B) brass connector
Connectors (Swagelok to female NPT)
Produktinformation
- TilgængelighedPrice
- HEWL0100-01181/8" (A)×1/4" (B) brass connector
- HEWL0100-01191/4" (A)×1/4" (B) brass connector
Reducing unions
Produktinformation
- TilgængelighedPrice
- HEWL0100-01211/8"×1/4" stainless steel reducing union
- HEWL5180-41301/16"×1/8" brass reducing unions
- HEWL5180-41311/8"×1/4" brass reducing unions
Reducers
Produktinformation
- TilgængelighedPrice
- HEWL5180-41341/4" (A)×1/8" (B) brass reducer
- HEWL5180-41351/8" (A)×1/4" (B) brass reducer
Fittings kit
Produktinformation
- TilgængelighedPrice
- HEWL5180-4161Fittings kit
Ferrule tools
Produktinformation
- TilgængelighedPrice
- HEWLRFT-5300Megabore ferrule tool, stainless steel, 0,53 mm, to remove ferrules stuck inside from fittingsMegabore ferrule tool, stainless steel, 0,53 mm, to remove ferrules stuck inside from fittingsLog ind for tilgængelighed1 * 1 Items284,00 kr
This product has been blocked by your organization. Please contact your purchasing department for more information