Extraction thimbles, borosilicate glass
Made from glass micro fibers without binder.
- Pollution monitoring, air sampling
- High temperature filtration (up to 500 °C)
- For solvents incompatible with cellulose thimbles (for example strong acids or bases)
Extraction thimbles for high temperatures and aggressive solvents
Made from glass micro fibers without binder.
- Pollution monitoring, air sampling
- High temperature filtration (up to 500 °C)
- For solvents incompatible with cellulose thimbles (for example strong acids or bases)
Extraction thimbles for high temperatures and aggressive solvents
Product Family Options
Grade MN 649
- Indv. Ø×HTykkelseModel
- 28×60 mm1,5 mmMN 649, for dust analysis in hot exhaust gases1 * 25 Items1.790,00 kr
- 15×50 mm1,0 mmMN 649, for dust analysis in hot exhaust gases1 * 25 Items1.790,00 kr
- 16×100 mm1,0 mmMN 649, for dust analysis in hot exhaust gases1 * 25 Items1.790,00 kr
- 22×80 mm1,5 mmMN 649, for dust analysis in hot exhaust gases1 * 25 Items1.790,00 kr
- 23×90 mm1,5 mmMN 649, for dust analysis in hot exhaust gases1 * 25 Items1.790,00 kr
- 23×100 mm1,5 mmMN 649, for dust analysis in hot exhaust gases1 * 25 Items1.790,00 kr
- 27×80 mm1,5 mmMN 649, for dust analysis in hot exhaust gases1 * 25 Items1.790,00 kr
- 28×120 mm1,5 mmMN 649, for dust analysis in hot exhaust gases1 * 25 Items2.130,00 kr
- 30×150 mm1,5 mmMN 649, for dust analysis in hot exhaust gases1 * 25 Items2.600,00 kr
- 33×90 mm1,5 mmMN 649, for dust analysis in hot exhaust gases1 * 25 Items2.160,00 kr
- 33×94 mm1,5 mmMN 649, for dust analysis in hot exhaust gases1 * 25 Items2.160,00 kr
- 33×100 mm1,5 mmMN 649, for dust analysis in hot exhaust gases1 * 25 Items2.160,00 kr
- 33×118 mm1,5 mmMN 649, for dust analysis in hot exhaust gases1 * 25 Items2.590,00 kr
- 33×205 mm1,5 mmMN 649, for dust analysis in hot exhaust gases1 * 25 Items3.470,00 kr
- 35×150 mm1,5 mmMN 649, for dust analysis in hot exhaust gases1 * 25 Items3.030,00 kr
- 43×123 mm2,0 mmMN 649, for dust analysis in hot exhaust gases1 * 25 Items3.280,00 kr
- 48×230 mm2,0 mmMN 649, for dust analysis in hot exhaust gases1 * 25 Items4.600,00 kr
- 57×315 mm2,0 mmMN 649, for dust analysis in hot exhaust gases1 * 25 Items7.130,00 kr
- 75×330 mm2,0 mmMN 649, for dust analysis in hot exhaust gases1 * 25 Items7.910,00 kr
- 33×80 mm1,5 mmMN 649, for dust analysis in hot exhaust gases1 * 25 Items1.790,00 kr
Grade MN 649 R
- Indv. Ø×HTykkelseModel
- 79×155 mmMN 649 R with collar, for dust analysis in industrial gases, exhaust gases and room air1 * 25 Items6.890,00 kr
- 27×55 mmMN 649 R with collar, for dust analysis in industrial gases, exhaust gases and room air1 * 25 Items2.320,00 kr