Orders LinkContactUsLinkComponent
VWR® Safe-Tamper System
VWR® Safe-Tamper System
Safe-Tamper System anti-tampering bag, 157×306 mm
Katalog # 720-1985
Leverandør:   VWR Collection
VWR® Safe-Tamper System
Katalog # 720-1985
Leverandør:  VWR Collection
VWR® Safe-Tamper System
VWR® Safe-Tamper System
Safe-Tamper System anti-tampering bag, 157×306 mm
Price
660,00 kr
Log ind for tilgængelighed
1 * 500 Items
Product Details & Documents
Anti-tamper disposable bags, in PE, with double pocket for internal hospital biological sample transportation. Double pocket bags: Two separate compartments for lab document and specimen. It is possible to customise the bags.
  • Transparent bags, yellow coloured to identify leakage of fluid and for a better visibility of the contents
  • Adhesive closure to prevent tampering and to preserve patient privacy
  • Printed biohazard logo and instructions
  • The bag is destroyed by opening it, preventing reuse and avoiding any risk of cross-contamination
Specifikationer
  • Beskrivelse:
    Safe-Tamper System anti-tampering bag
  • L×B:
    157×306 mm