Phase separating paper
Om denne vare
These papers are made hydrophobic (impermeable to water) by impregnation with a silicone. With the aid of these filters, water can be separated from water-immiscible organic solvents in an elegant manner, by means of a simple filtration.
22 tilgængelige muligheder nedenfor
Product Details & Documents
These papers are made hydrophobic (impermeable to water) by impregnation with a silicone. With the aid of these filters, water can be separated from water-immiscible organic solvents in an elegant manner, by means of a simple filtration.
Specifications for Product Family
Specifikationer
- Catalog No:
- MANA430009
- MANA430011
- MANA430012
- MANA430015
- MANA430018
- MANA430024
- MANA484011
- MANA484015
- MANA484005
- MANA484007
- MANA484009
- MANA484012
- MANA484018
- MANA484024
- MANA484032
- MANA584011
- 516-6012
- 516-6019
- MANA584009
- MANA584018
- MANA584024
- MANA584032
- Vægt:
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- Tykkelse:
- 0,2 mm
- 0,2 mm
- 0,2 mm
- 0,2 mm
- 0,2 mm
- 0,2 mm
- 0,2 mm
- 0,2 mm
- 0,2 mm
- 0,2 mm
- 0,2 mm
- 0,2 mm
- 0,2 mm
- 0,2 mm
- 0,2 mm
- 0,2 mm
- 0,2 mm
- 0,2 mm
- 0,2 mm
- 0,2 mm
- 0,2 mm
- 0,2 mm
- Filtreringshastighed:
- 8 s (Herzberg)
- 8 s (Herzberg)
- 8 s (Herzberg)
- 8 s (Herzberg)
- 8 s (Herzberg)
- 8 s (Herzberg)
- 27 s (Herzberg)
- 27 s (Herzberg)
- 27 s (Herzberg)
- 27 s (Herzberg)
- 27 s (Herzberg)
- 27 s (Herzberg)
- 27 s (Herzberg)
- 27 s (Herzberg)
- 27 s (Herzberg)
- 27 s (Herzberg)
- 27 s (Herzberg)
- 27 s (Herzberg)
- 27 s (Herzberg)
- 27 s (Herzberg)
- 27 s (Herzberg)
- 27 s (Herzberg)
Specifikationer
Product Family Options
Grade MN 617 WA
Produktinformation
- KvalitetStørrelseTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.MANA430009Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed8 s (Herzberg) - Specifikationer:
More
Specifications
Cat. No.MANA430011Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed8 s (Herzberg) - Specifikationer:
More
Specifications
Cat. No.MANA430012Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed8 s (Herzberg) - Specifikationer:
More
Specifications
Cat. No.MANA430015Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed8 s (Herzberg) - Specifikationer:
More
Specifications
Cat. No.MANA430018Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed8 s (Herzberg) - Specifikationer:
More
Specifications
Cat. No.MANA430024Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed8 s (Herzberg)
Grade MN 616 WA
Produktinformation
- KvalitetStørrelseTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.MANA484011Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed27 s (Herzberg) - Specifikationer:
More
Specifications
Cat. No.MANA484015Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed27 s (Herzberg) - Specifikationer:
More
Specifications
Cat. No.MANA484005Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed27 s (Herzberg) - Specifikationer:
More
Specifications
Cat. No.MANA484007Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed27 s (Herzberg) - Specifikationer:
More
Specifications
Cat. No.MANA484009Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed27 s (Herzberg) - Specifikationer:
More
Specifications
Cat. No.MANA484012Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed27 s (Herzberg) - Specifikationer:
More
Specifications
Cat. No.MANA484018Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed27 s (Herzberg) - Specifikationer:
More
Specifications
Cat. No.MANA484024Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed27 s (Herzberg) - Specifikationer:
More
Specifications
Cat. No.MANA484032Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed27 s (Herzberg)
Grade MN 616 WA, folded
Produktinformation
- KvalitetStørrelseTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.MANA584011Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed27 s (Herzberg) - Specifikationer:
More
Specifications
Cat. No.516-6012Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed27 s (Herzberg) - Specifikationer:
More
Specifications
Cat. No.516-6019Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed27 s (Herzberg) - Specifikationer:
More
Specifications
Cat. No.MANA584009Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed27 s (Herzberg) - Specifikationer:
More
Specifications
Cat. No.MANA584018Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed27 s (Herzberg) - Specifikationer:
More
Specifications
Cat. No.MANA584024Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed27 s (Herzberg) - Specifikationer:
More
Specifications
Cat. No.MANA584032Vægt85 g/m²Tykkelse0,2 mmFiltreringshastighed27 s (Herzberg)