Orders LinkContactUsLinkComponent
Handheld magnifiers
Handheld magnifiers
2,3×, 80 mm
Katalog # 630-8156
Leverandør:   ESCHENBACH OPTIK
Handheld magnifiers
Katalog # 630-8156
Leverandør:  ESCHENBACH OPTIK
Handheld magnifiers
Handheld magnifiers
2,3×, 80 mm
Price
408,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Handheld magnifiers with biconvex lens.
  • Lens for a distortion free image with edge-to-edge sharpness
  • Transparent plastic frames
  • Excellent orientation on the reading material
Specifikationer
  • Ø:
    80 mm
  • Forstørrelse:
    2,3×
  • Seq:
    02