Solid phase extraction cartridges, Bond Elut™ SI
Om denne vare
Native silica is generally regarded as the most polar SPE sorbent available. Bond Elut™ SI is particularly effective at separating compounds with a very similar structure. Applying the analytes in a non-polar solvent, then increasing the solvent polarity by increasing the concentration of a polar modifier, such as THF or ethyl acetate, delivers effective separations.
- Highly polar phase retains polar molecules from non-polar matrices
- High purity silica
- Separate compounds with very similar structures
18 tilgængelige muligheder nedenfor
Product Details & Documents
Native silica is generally regarded as the most polar SPE sorbent available. Bond Elut™ SI is particularly effective at separating compounds with a very similar structure. Applying the analytes in a non-polar solvent, then increasing the solvent polarity by increasing the concentration of a polar modifier, such as THF or ethyl acetate, delivers effective separations.
- Highly polar phase retains polar molecules from non-polar matrices
- High purity silica
- Separate compounds with very similar structures
Specifications for Product Family
Specifikationer
- Catalog No:
- VARI12113010
- VARI14113036
- 551-0346
- VARI14113010
- 549-0789
- 551-0306
- 551-0321
- 551-0407
- 549-0803
- 549-0851
- 549-0841
- 549-0846
- 549-0856
- 549-0861
- 549-0866
- 549-0870
- 551-0375
- 551-0382
- Endcapped:
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Surface area:
- 600 m²/g
- 500 m²/g
- 600 m²/g
- 600 m²/g
- 600 m²/g
- 600 m²/g
- 600 m²/g
- 600 m²/g
- 600 m²/g
- 600 m²/g
- 600 m²/g
- 600 m²/g
- 600 m²/g
- 600 m²/g
- 600 m²/g
- 600 m²/g
- 600 m²/g
Specifikationer
Product Family Options
LRC cartridges
Produktinformation
- FasePorestørrelsePartikelstørrelseKapacitetLejevægtTilgængelighedPrice
- VARI12113010LRC cartridgeSpecifikationer:
More
Specifications
Cat. No.VARI12113010EndcappedIngenSurface area600 m²/g - Specifikationer:
More
Specifications
Cat. No.VARI14113036EndcappedSurface area - 551-0346LRC-patronerSpecifikationer:
More
Specifications
Cat. No.551-0346EndcappedIngenSurface area500 m²/g - VARI14113010LRC cartridgeSpecifikationer:
More
Specifications
Cat. No.VARI14113010EndcappedIngenSurface area600 m²/g
Straight barrel cartridges
Produktinformation
- FasePorestørrelsePartikelstørrelseKapacitetLejevægtTilgængelighedPrice
- 549-0789Straight barrel cartridgeStraight barrel cartridgeSilica60 Å40 µm1 ml50 mgLog ind for tilgængelighed1 * 100 Items1.900,00 krSpecifikationer:
More
Specifications
Cat. No.549-0789EndcappedIngenSurface area600 m²/g - 551-0306Straight barrel cartridgeSpecifikationer:
More
Specifications
Cat. No.551-0306EndcappedIngenSurface area600 m²/g - 551-0321Straight barrel cartridgeStraight barrel cartridgeSilica60 Å40 µm3 ml500 mgLog ind for tilgængelighed1 * 50 Items1.800,00 krSpecifikationer:
More
Specifications
Cat. No.551-0321EndcappedIngenSurface area600 m²/g - 551-0407Straight barrel cartridgeStraight barrel cartridgeSilica60 Å40 µm6 ml1000 mgLog ind for tilgængelighed1 * 30 Items1.500,00 krSpecifikationer:
More
Specifications
Cat. No.551-0407EndcappedIngenSurface area600 m²/g - 549-0803Straight barrel cartridgeStraight barrel cartridgeSilica60 Å40 µm3 ml1500 mgLog ind for tilgængelighed1 * 50 Items2.240,00 krSpecifikationer:
More
Specifications
Cat. No.549-0803EndcappedIngenSurface area600 m²/g - 549-0851Straight barrel cartridgeStraight barrel cartridgeSilica60 Å120 µm1 ml50 mgLog ind for tilgængelighed1 * 100 Items1.900,00 krSpecifikationer:
More
Specifications
Cat. No.549-0851EndcappedIngenSurface area600 m²/g - 549-0841Straight barrel cartridgeStraight barrel cartridgeSilica60 Å120 µm1 ml100 mgLog ind for tilgængelighed1 * 100 Items2.070,00 krSpecifikationer:
More
Specifications
Cat. No.549-0841EndcappedIngenSurface area600 m²/g - 549-0846Straight barrel cartridgeStraight barrel cartridgeSilica60 Å120 µm3 ml500 mgLog ind for tilgængelighed1 * 50 Items1.900,00 krSpecifikationer:
More
Specifications
Cat. No.549-0846EndcappedIngenSurface area600 m²/g - 549-0856Straight barrel cartridgeStraight barrel cartridgeSilica60 Å120 µm6 ml1000 mgLog ind for tilgængelighed1 * 30 Items1.500,00 krSpecifikationer:
More
Specifications
Cat. No.549-0856EndcappedIngenSurface area600 m²/g - 549-0861Straight barrel cartridgeStraight barrel cartridgeSilica60 Å120 µm6 ml2000 mgLog ind for tilgængelighed1 * 20 Items1.720,00 krSpecifikationer:
More
Specifications
Cat. No.549-0861EndcappedIngenSurface area600 m²/g - 549-0866Straight barrel cartridgeStraight barrel cartridgeSilica60 Å120 µm20 ml5000 mgLog ind for tilgængelighed1 * 20 Items2.090,00 krSpecifikationer:
More
Specifications
Cat. No.549-0866EndcappedIngenSurface area600 m²/g - 549-0870Straight barrel cartridgeStraight barrel cartridgeSilica60 Å120 µm60 ml10000 mgLog ind for tilgængelighed1 * 16 Items2.420,00 krSpecifikationer:
More
Specifications
Cat. No.549-0870EndcappedIngenSurface area600 m²/g
Bond Elut™ Jr