Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ PSA
Solid phase extraction cartridges, Bond Elut™ PSA
Leverandør:   VARIAN
Solid phase extraction cartridges, Bond Elut™ PSA
Leverandør:  VARIAN
Certifikater
Solid phase extraction cartridges, Bond Elut™ PSA
8 tilgængelige muligheder nedenfor
Product Details & Documents
Bond Elut™ PSA is an alkylated amine sorbent that contains two different amino functionalities, one secondary and one primary. This gives a slightly higher pKa and ionic capacity compared to Bond Elut™ NH₂. The PSA functional group is a very effective bidentate ligand and is therefore an excellent sorbent for chelation applications. PSA has a significantly higher carbon load than most amino functional sorbents and thus is a better choice for polar compounds, which retain too strongly on Bond Elut™ NH₂.
  • Very effective bidentate ligand
  • Alternative choice for polar compounds to Bond Elut™ NH₂
  • Higher ionic capacity than NH₂
Specifications for Product Family
Specifikationer
  • Catalog No:
  • VARI12113041
  • VARI12102077
  • VARI12102015
  • VARI12102042
  • VARI12256140
  • VARI12256055
  • 551-0378
  • VARI12166050B
  • Kulstofbelastning:
  • 7,5%
  • 7,5%
  • 7,5%
  • 7,5%
  • 7,5%
  • 7,5%
  • 7,5%
  • 7,5%
  • Endcapped:
  • Ingen
  • Ingen
  • Ingen
  • Ingen
  • Ingen
  • Ingen
  • Ingen
  • Ingen
  • Surface area:
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
Specifikationer
Product Family Options
LRC cartridges
Produktinformation
  • Fase
    Porestørrelse
    Partikelstørrelse
    Kapacitet
    Lejevægt
    Tilgængelighed
    Price
  • VARI12113041
    LRC-patroner
    LRC-patroner
    PSA
    60 Å
    10 ml
    500 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 50 Items
    1 * 50 Items - 2.580,00 kr
    2.580,00 kr
    1 * 50 Items
       2.580,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    VARI12113041
    Kulstofbelastning
    7,5%
    Endcapped
    Ingen
    Surface area
    500 m²/g
Straight barrel cartridges
Produktinformation
  • Fase
    Porestørrelse
    Partikelstørrelse
    Kapacitet
    Lejevægt
    Tilgængelighed
    Price
  • VARI12102077
    Straight barrel cartridge
    Straight barrel cartridge
    PSA
    60 Å
    40 µm
    1 ml
    50 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 3.120,00 kr
    3.120,00 kr
    1 * 100 Items
       3.120,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    VARI12102077
    Kulstofbelastning
    7,5%
    Endcapped
    Ingen
    Surface area
    500 m²/g
  • VARI12102015
    Straight barrel cartridge
    Straight barrel cartridge
    PSA
    60 Å
    40 µm
    1 ml
    100 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items
    Specifikationer:
    More
    Specifications
    Cat. No.
    VARI12102015
    Kulstofbelastning
    7,5%
    Endcapped
    Ingen
    Surface area
    500 m²/g
  • VARI12102042
    Straight barrel cartridge
    Straight barrel cartridge
    PSA
    60 Å
    120 µm
    3 ml
    500 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 50 Items
    1 * 50 Items - 2.730,00 kr
    2.730,00 kr
    1 * 50 Items
       2.730,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    VARI12102042
    Kulstofbelastning
    7,5%
    Endcapped
    Ingen
    Surface area
    500 m²/g
  • VARI12256140
    Straight barrel cartridge
    Straight barrel cartridge
    PSA
    60 Å
    40 µm
    6 ml
    1 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 30 Items
    1 * 30 Items - 2.310,00 kr
    2.310,00 kr
    1 * 30 Items
       2.310,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    VARI12256140
    Kulstofbelastning
    7,5%
    Endcapped
    Ingen
    Surface area
    500 m²/g
  • VARI12256055
    Straight barrel cartridge
    Straight barrel cartridge
    PSA
    60 Å
    40 µm
    12 ml
    2 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 20 Items
    1 * 20 Items - 2.460,00 kr
    2.460,00 kr
    1 * 20 Items
       2.460,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    VARI12256055
    Kulstofbelastning
    7,5%
    Endcapped
    Ingen
    Surface area
    500 m²/g
Bond Elut™ Jr
Produktinformation
  • Fase
    Porestørrelse
    Partikelstørrelse
    Kapacitet
    Lejevægt
    Tilgængelighed
    Price
  • 551-0378
    Bond Elut™ Jr
    Bond Elut™ Jr
    PSA
    60 Å
    500 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 3.490,00 kr
    3.490,00 kr
    1 * 100 Items
       3.490,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    551-0378
    Kulstofbelastning
    7,5%
    Endcapped
    Ingen
    Surface area
    500 m²/g
  • VARI12166050B
    Bond Elut™ Jr
    Bond Elut™ Jr
    PSA
    60 Å
    1 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 4.170,00 kr
    4.170,00 kr
    1 * 100 Items
       4.170,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    VARI12166050B
    Kulstofbelastning
    7,5%
    Endcapped
    Ingen
    Surface area
    500 m²/g