Solid phase extraction cartridges, Bond Elut™ NH₂
Om denne vare
Bond Elut™ NH₂ is a weaker anion exchanger than sorbents such as SAX (a quaternary amine sorbent that is always charged) and is therefore a better choice for retention of very strong anions, such as sulfonic acids, which may retain irreversibly on a SAX sorbent. Similar to Diol and SI sorbents, Bond Elut™ NH₂ is excellent for the separation of structural isomers.
- Weaker anion exchange than SAX
- Very good for separating structural isomers
- Uses hydrogen bonding and anion exchange
24 tilgængelige muligheder nedenfor
Product Details & Documents
Bond Elut™ NH₂ is a weaker anion exchanger than sorbents such as SAX (a quaternary amine sorbent that is always charged) and is therefore a better choice for retention of very strong anions, such as sulfonic acids, which may retain irreversibly on a SAX sorbent. Similar to Diol and SI sorbents, Bond Elut™ NH₂ is excellent for the separation of structural isomers.
- Weaker anion exchange than SAX
- Very good for separating structural isomers
- Uses hydrogen bonding and anion exchange
Specifications for Product Family
Specifikationer
- Catalog No:
- VARI12113014
- VARI12113067
- VARI12113040
- VARI14113040
- 549-0792
- 551-0310
- 549-0795
- 549-0799
- 549-0801
- 551-0324
- 549-0831
- 549-0800
- 549-0822
- 549-0853
- 549-0858
- 549-0863
- VARI12102160
- 551-0377
- 551-0384
- VARI75405050
- 551-0430
- VARI75505050
- VARI7550501C
- VARI7553502C
- Kulstofbelastning:
- 6,7%
- 6,7%
- 6,7%
- 6,7%
- 6,7%
- 6,7%
- 6,7%
- 6,7%
- 6,7%
- 6,7%
- 6,7%
- 6,7%
- 6,7%
- 6,7%
- 6,7%
- 6,7%
- 6,7%
- 6,7%
- 6,7%
- 6,7%
- 6,7%
- 6,7%
- 6,7%
- 6,7%
- Endcapped:
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Surface area:
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
Specifikationer
Product Family Options
LRC cartridges
Produktinformation
- FasePorestørrelsePartikelstørrelseKapacitetLejevægtTilgængelighedPrice
- VARI12113014LRC cartridgeSpecifikationer:
More
Specifications
Cat. No.VARI12113014Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g - VARI12113067LRC cartridgeSpecifikationer:
More
Specifications
Cat. No.VARI12113067Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g - VARI12113040LRC cartridgeSpecifikationer:
More
Specifications
Cat. No.VARI12113040Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g - VARI14113040LRC cartridgeSpecifikationer:
More
Specifications
Cat. No.VARI14113040Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g
Straight barrel cartridges
Produktinformation
- FasePorestørrelsePartikelstørrelseKapacitetLejevægtTilgængelighedPrice
- 549-0792Straight barrel cartridgeSpecifikationer:
More
Specifications
Cat. No.549-0792Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g - 551-0310Straight barrel cartridgeSpecifikationer:
More
Specifications
Cat. No.551-0310Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g - 549-0795Straight barrel cartridgeSpecifikationer:
More
Specifications
Cat. No.549-0795Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g - 549-0799Straight barrel cartridgeSpecifikationer:
More
Specifications
Cat. No.549-0799Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g - 549-0801Straight barrel cartridgeSpecifikationer:
More
Specifications
Cat. No.549-0801Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g - 551-0324Straight barrel cartridgeSpecifikationer:
More
Specifications
Cat. No.551-0324Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g - 549-0831Straight barrel cartridgeSpecifikationer:
More
Specifications
Cat. No.549-0831Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g - 549-0800Straight barrel cartridgeSpecifikationer:
More
Specifications
Cat. No.549-0800Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g - 549-0822Straight barrel cartridgeSpecifikationer:
More
Specifications
Cat. No.549-0822Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g - 549-0853Straight barrel cartridgeSpecifikationer:
More
Specifications
Cat. No.549-0853Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g - 549-0858Straight barrel cartridgeSpecifikationer:
More
Specifications
Cat. No.549-0858Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g - 549-0863Straight barrel cartridgeSpecifikationer:
More
Specifications
Cat. No.549-0863Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g - VARI12102160Straight barrel cartridgeSpecifikationer:
More
Specifications
Cat. No.VARI12102160Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g
Bond Elut™ Jr
Produktinformation
- FasePorestørrelsePartikelstørrelseKapacitetLejevægtTilgængelighedPrice
- 551-0377Bond Elut™ JrSpecifikationer:
More
Specifications
Cat. No.551-0377Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g - 551-0384Bond Elut™ JrSpecifikationer:
More
Specifications
Cat. No.551-0384Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g
VersaPlate™ formats
Produktinformation
- FasePorestørrelsePartikelstørrelseKapacitetLejevægtTilgængelighedPrice
- VARI75405050Formonteret 96-brønds pladeSpecifikationer:
More
Specifications
Cat. No.VARI75405050Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g - 551-0430Formonteret 96-brønds pladeSpecifikationer:
More
Specifications
Cat. No.551-0430Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g - VARI75505050Versaplate™-rørSpecifikationer:
More
Specifications
Cat. No.VARI75505050Kulstofbelastning6,7%EndcappedIngenSurface area500 m²/g - VARI7550501CVersaplate™-rørSpecifikationer:
More
Specifications
Cat. No.VARI7550501CKulstofbelastning6,7%EndcappedIngenSurface area500 m²/g - VARI7553502CVersaPlate™ tubeSpecifikationer:
More
Specifications
Cat. No.VARI7553502CKulstofbelastning6,7%EndcappedIngenSurface area500 m²/g