2'-Hydroxyflavanone ≥98.0% (by HPLC)
Glass bottle, 1 g
Katalog # TCIAH1024-1G
Leverandør: TCI
Gå til:
Product Details & Documents
- CAS nummer:17348-76-4
- Formel:C₁₅H₁₂O₃
- MDL nummer:MFCD00017702
- Molecular Weight:240.25 g/mol
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:164.0 to 168.0 deg-C
Specifikationer
- Pakning:Glass bottle
- Pakn:1 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



