Methyldecanoat
Glass bottle, 100 mL
Katalog # ROTH3325.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Decanoic acid methyl ester, Capric acid methyl ester, Decylic acid methyl ester, Methyl caprate
- CAS nummer:110-42-9
- Formel:C₁₁H₂₂O₂
- MDL nummer:MFCD00009580
- Molecular Weight:186.29 g/mol
- Kogende Pt:224 °C (1013 hPa)
- Smeltende Pt:–18 °C
- Tæthed:0,8724 g/cm³ (20 °C)
- Flash Pt:94 °C
- EINECS:203-766-6
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:100 mL