Solid phase extraction cartridges, Bond Elut™ CN-E
Leverandør: VARIAN
Om denne vare
A medium polarity sorbent with many uses, Bond Elut™ CN-E is ideal for applications in which extremely non-polar compounds would be irreversibly retained on high carbon load sorbents such as C₈ and C₁₈. This endcapped version of the cyano sorbent is best utilised when extracting analytes from an aqueous matrix.
- Ideal for extracting aqueous analytes
- Retention in aqueous and organic matrices
- Useful for many applications
4 tilgængelige muligheder nedenfor
Product Details & Documents
A medium polarity sorbent with many uses, Bond Elut™ CN-E is ideal for applications in which extremely non-polar compounds would be irreversibly retained on high carbon load sorbents such as C₈ and C₁₈. This endcapped version of the cyano sorbent is best utilised when extracting analytes from an aqueous matrix.
- Ideal for extracting aqueous analytes
- Retention in aqueous and organic matrices
- Useful for many applications
Specifications for Product Family
Specifikationer
- Catalog No:
- VARI12113033
- VARI12102064
- VARI12102007
- VARI12102034
- Kulstofbelastning:
- 8,1%
- 8,1%
- 8,1%
- 8,1%
- Endcapped:
- Ja
- Ja
- Ja
- Ja
- Surface area:
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
Specifikationer
Product Family Options
LRC cartridges
Produktinformation
- FaseKapacitetLejevægtTilgængelighedPrice
- VARI12113033LRC-patronerSpecifikationer:
More
Specifications
Cat. No.VARI12113033Kulstofbelastning8,1%EndcappedJaSurface area500 m²/g
Straight barrel cartridges
Produktinformation
- FaseKapacitetLejevægtTilgængelighedPrice
- VARI12102064Straight barrel cartridgeSpecifikationer:
More
Specifications
Cat. No.VARI12102064Kulstofbelastning8,1%EndcappedJaSurface area500 m²/g - VARI12102007Straight barrel cartridgeSpecifikationer:
More
Specifications
Cat. No.VARI12102007Kulstofbelastning8,1%EndcappedJaSurface area500 m²/g - VARI12102034Straight barrel cartridgeSpecifikationer:
More
Specifications
Cat. No.VARI12102034Kulstofbelastning8,1%EndcappedJaSurface area500 m²/g