Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ CBA
Solid phase extraction cartridges, Bond Elut™ CBA
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ CBA
Supplier:  VARIAN
Certificates
Solid phase extraction cartridges, Bond Elut™ CBA
8 Options Available Below
Product Details & Documents
CBA is a mid-polarity sorbent and weak cation exchanger (pKa 4,8). It can be used with a wider range of counter-ions than lower pKa sorbents like SCX and will demonstrate easier elution of quaternary amine functionalised analytes.
  • Cation exchange with no need for extreme basic conditions
  • Wider selectivity range provides more eluent options
  • Polar or non-polar depending on matrix or solvent
Specifications for Product Family
Specifications
  • Catalog No:
  • VARI12113011
  • VARI12113037
  • VARI12102073
  • 551-0307
  • VARI12102097
  • VARI12102124
  • VARI12256009
  • VARI12256058
  • Kulstofbelastning:
  • 7,4%
  • 7,4%
  • 7,4%
  • 7,4%
  • 7,4%
  • 7,4%
  • 7,4%
  • 7,4%
  • Endcapped:
  • Ja
  • Ja
  • Ja
  • Ja
  • Ja
  • Ja
  • Ja
  • Ja
  • Surface area:
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
Specifications
Product Family Options
LRC cartridges
Product Information
  • Beskrivelse
    Fase
    Porestørrelse
    Kapacitet
    Lejevægt
    Availability
    Price
  • LRC-patroner
    CBA
    60 Å
    10 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - 1.880,00 kr
    1.880,00 kr
    1 * 50 Items
       1.880,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12113011
    Kulstofbelastning
    7,4%
    Endcapped
    Ja
    Surface area
    500 m²/g
  • LRC-patroner
    CBA
    60 Å
    10 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - 2.640,00 kr
    2.640,00 kr
    1 * 50 Items
       2.640,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12113037
    Kulstofbelastning
    7,4%
    Endcapped
    Ja
    Surface area
    500 m²/g
Straight barrel cartridges
Product Information
  • Beskrivelse
    Fase
    Porestørrelse
    Kapacitet
    Lejevægt
    Availability
    Price
  • Straight barrel cartridge
    CBA
    60 Å
    1 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 3.380,00 kr
    3.380,00 kr
    1 * 100 Items
       3.380,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102073
    Kulstofbelastning
    7,4%
    Endcapped
    Ja
    Surface area
    500 m²/g
  • Straight barrel cartridge
    CBA
    60 Å
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items
    Specifications:
    More
    Specifications
    Cat. No.
    551-0307
    Kulstofbelastning
    7,4%
    Endcapped
    Ja
    Surface area
    500 m²/g
  • Straight barrel cartridge
    CBA
    60 Å
    3 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - 2.440,00 kr
    2.440,00 kr
    1 * 50 Items
       2.440,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102097
    Kulstofbelastning
    7,4%
    Endcapped
    Ja
    Surface area
    500 m²/g
  • Straight barrel cartridge
    CBA
    60 Å
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - 2.250,00 kr
    2.250,00 kr
    1 * 50 Items
       2.250,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102124
    Kulstofbelastning
    7,4%
    Endcapped
    Ja
    Surface area
    500 m²/g
  • Straight barrel cartridge
    CBA
    60 Å
    6 ml
    1 g
    Log in for availability
    Log in for availability
    1 * 30 Items
    1 * 30 Items - 2.310,00 kr
    2.310,00 kr
    1 * 30 Items
       2.310,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12256009
    Kulstofbelastning
    7,4%
    Endcapped
    Ja
    Surface area
    500 m²/g
  • Straight barrel cartridge
    CBA
    60 Å
    12 ml
    2 g
    Log in for availability
    Log in for availability
    1 * 20 Items
    1 * 20 Items - 2.010,00 kr
    2.010,00 kr
    1 * 20 Items
       2.010,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12256058
    Kulstofbelastning
    7,4%
    Endcapped
    Ja
    Surface area
    500 m²/g