Bunsen burner
Bunsen burner with air regulation, All gases, 17 mm
Katalog # HECH41385010
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
All gas bunsen burner with air regulation, needle valve and adjustable nozzle.
- Height of approx. 160 mm
- Tube Ø of approx. 13 mm
Specifikationer
- Beskrivelse:Bunsen burner with air regulation
- Ø hoved:17 mm
- Til:All gases