Solid phase extraction cartridges, Bond Elut™ C18 OH
Bond Elut™ C18 OH is a non-end capped version of the octadecyl bonded phases that enables the silanols on the silica surface to be more active. This low-load C18 has well-controlled silanol activity that permits the fractionation of metabolites and enhances retention of basic compounds compared to an endcapped C18. The pore size makes it a good choice for peptide extraction.
- Silanol activity permits metabolite fractionation
- Tight QC tolerances deliver batch-to-batch reproducibility
- 150 Å pore size expands retention window to higher molecular weight compounds
Bond Elut™ C18 OH is a non-end capped version of the octadecyl bonded phases that enables the silanols on the silica surface to be more active. This low-load C18 has well-controlled silanol activity that permits the fractionation of metabolites and enhances retention of basic compounds compared to an endcapped C18. The pore size makes it a good choice for peptide extraction.
- Silanol activity permits metabolite fractionation
- Tight QC tolerances deliver batch-to-batch reproducibility
- 150 Å pore size expands retention window to higher molecular weight compounds
- Catalog No:
- VARI12256040
- VARI12102065
- VARI12102020
- VARI12102046
- Kulstofbelastning:
- 14,9%
- 14,9%
- 14,9%
- 14,9%
- Endcapped:
- Ingen
- Ingen
- Ingen
- Ingen
- Surface area:
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
Product Family Options
- BeskrivelseFasePorestørrelsePartikelstørrelseKapacitetLejevægt
- Lige cylinderpatronC18 OH150 Å40 and 1206 ml1 g1 * 30 Items1.910,00 kr
Specifications
Cat. No.VARI12256040Kulstofbelastning14,9%EndcappedIngenSurface area300 m²/g - Straight barrel cartridgeC18 OH150 Å1 ml50 mg1 * 100 Items2.490,00 kr
Specifications
Cat. No.VARI12102065Kulstofbelastning14,9%EndcappedIngenSurface area300 m²/g Specifications
Cat. No.VARI12102020Kulstofbelastning14,9%EndcappedIngenSurface area300 m²/g- Lige cylinderpatronC18 OH150 Å3 ml500 mg1 * 50 Items2.060,00 kr
Specifications
Cat. No.VARI12102046Kulstofbelastning14,9%EndcappedIngenSurface area300 m²/g