Orders LinkContactUsLinkComponent
Automatic burettes, Dr Schilling pattern, Schellbach stripe
Automatic burettes, Dr Schilling pattern, Schellbach stripe
Automatic burettes, Dr Schilling pattern, Schellbach stripe
Automatic burettes, Dr Schilling pattern, Schellbach stripe
AR-GLAS®, class B.
  • Burettes are calibrated to deliver (marked 'Ex')
  • Schellbach stripe and fused graduation, faceted outlet tip
  • Improved special pinch cock with titration screw for precise regulation
Precisely calibrated, tolerance 1,5 times the official tolerance of class AS
Product Family Options
  • Kapacitet
    Inddeling
    Tolerance
  • 5 ml
    0,05 ml
    0,050 ± ml
    1 * 1 Items
    1 * 1 Items - 661,00 kr
    661,00 kr
    1 * 1 Items
       661,00 kr
  • 10 ml
    0,05 ml
    0,050 ± ml
    1 * 1 Items
    1 * 1 Items - 701,00 kr
    701,00 kr
    1 * 1 Items
       701,00 kr
  • 15 ml
    0,1 ml
    0,100 ± ml
    1 * 1 Items
    1 * 1 Items - 669,00 kr
    669,00 kr
    1 * 1 Items
       669,00 kr
  • 50 ml
    0,1 ml
    0,100 ± ml
    1 * 1 Items
    1 * 1 Items - 780,00 kr
    780,00 kr
    1 * 1 Items
       780,00 kr
Accessories
  •  
    Beskrivelse
    Kapacitet
    Inddeling
  • HECH40821005
    Ekstra burette, AR®/sodaglas
    5 ml
    0,05 ml
    1 * 1 Items - 400,00 kr
  •  
    Beskrivelse
    Kapacitet
    Inddeling
  • HECH40823001
    Ekstra plastikbase, størrelse 1, til 500 ml Schilling-flasker
    -
    -
    1 * 1 Items - 47,80 kr